
3-Methoxy-3-methylbutanol

Synonym(s):
3-Methoxy-3-methyl-1-butanol
Linear Formula:
(CH3)2C(OCH3)CH2CH2OH
CAS Number:
Molecular Weight:
118.17
Products
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing