Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3-Hydroxy-4-methoxyphenethylamine hydrochloride
Synonym(s):
4-Methoxytyramine hydrochloride, 4-O-Methyldopamine hydrochloride
Empirical Formula (Hill Notation):
C
9
H
13
NO
2
· HCl
CAS Number:
645-33-0
Molecular Weight:
203.67
EC Number:
211-437-3
Products
Product Category
bioactive small molecules (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Research Area
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
H3132
3-Hydroxy-4-methoxyphenethylamine hydrochloride,
Expand
View Pricing