Skip to Content
Merck
2-Pentadecanone ≥98%, FG

2-Pentadecanone

2-Pentadecanone ≥98%, FG
Synonym(s):
Methyl tridecyl ketone
Linear Formula:
CH3CO(CH2)12CH3
CAS Number:
Molecular Weight:
226.40
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing