
2-Methoxyestradiol

Synonym(s):
1,3,5(10)-Estratriene-2,3,17-triol 2-methyl ether, 2,3,17β-Trihydroxy-1,3,5(10)-estratriene 2-methyl ether, 2-Hydroxyestradiol 2-methyl ether, 3,17β-Dihydroxy-2-methoxy-1,3,5(10)-estratriene, 2-Methoxyestradiol
Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Special Grade
Sterility
Showing 1-2 of 2
Compare
Product Number
Description
Pricing