Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Imidazolidinethione
Synonym(s):
2-Thioxoimidazolidine,
N,N′
-Ethylenethiourea, 2-Imidazolidinethione
Empirical Formula (Hill Notation):
C
3
H
6
N
2
S
CAS Number:
96-45-7
Molecular Weight:
102.16
EC Number:
202-506-9
Products
Product Category
building blocks (2)
pesticides (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Purum (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
I504
2-Imidazolidinethione,
98%
Expand
View Pricing
8.20696
2-Imidazolidinethione,
for synthesis
Expand
View Pricing
45531
2-Imidazolidinethione,
PESTANAL
®
, analytical standard
Expand
View Pricing
03940
2-Imidazolidinethione,
purum, ≥98.0% (HPLC)
Expand
View Pricing