Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Aminopyridine
Synonym(s):
2-AP, 2-Pyridinamine, 2-Pyridylamine, 2-Aminopyridine
Linear Formula:
2-(NH
2
)C
5
H
4
N
CAS Number:
504-29-0
Molecular Weight:
94.11
EC Number:
207-988-4
Products
Product Category
building blocks (3)
pesticides (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Purum (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
A77989
2-Aminopyridine,
≥99%
Expand
View Pricing
A77997
2-Aminopyridine,
99%
Expand
View Pricing
8.01113
2-Aminopyridine,
for synthesis
Expand
View Pricing
09340
2-Aminopyridine,
purum, ≥98.0% (NT)
Expand
View Pricing
36685
2-Aminopyridine,
PESTANAL
®
, analytical standard
Expand
View Pricing