
1H,1H,2H,2H-Perfluorooctanesulfonic acid

Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanesulfonic acid, 1H,1H′,2H,2H′-Perfluorooctane sulfonic acid, 1H,1H,2H,2H-Perfluoro-1-octanesulfonic acid, 6:2 Fluorotelomer sulfonic acid
Empirical Formula (Hill Notation):
C8H5F13O3S
CAS Number:
Molecular Weight:
428.17
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing