![1-Ethyl-3-methylimidazolium acetate ≥98%](/deepweb/assets/sigmaaldrich/product/structures/117/598/c2889391-2d44-4876-92f5-fca61b3871e6/640/c2889391-2d44-4876-92f5-fca61b3871e6.png)
1-Ethyl-3-methylimidazolium acetate
![1-Ethyl-3-methylimidazolium acetate ≥98%](/deepweb/assets/sigmaaldrich/product/structures/117/598/c2889391-2d44-4876-92f5-fca61b3871e6/640/c2889391-2d44-4876-92f5-fca61b3871e6.png)
Synonym(s):
EMIM Ac
Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Geographic Origin
Application
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing