Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,2:3,4-Di-
O
-isopropylidene-α-D-galactopyranose
Synonym(s):
1,2:3,4-Di-
O
-isopropylidene-D-galactopyranose
Empirical Formula (Hill Notation):
C
12
H
20
O
6
CAS Number:
4064-06-6
Molecular Weight:
260.28
EC Number:
223-771-7
Products
Product Category
building blocks (1)
carbohydrates (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D126306
1,2:3,4-Di-
O
-isopropylidene-α-D-galactopyranose,
97%
Expand
View Pricing