Merck
Membrane Chromatography
Our Natrix® chromatography membrane technology provides a new level of downstream productivity and versatility. The macroporous structure, combined with high functional group density, enables high load capacity and excellent impurity removal at ultra-fast flow rates. With great performance in a single-use and scalable format, our innovative Natrix® products improve process economics and facility utilization.
Products
Compare
Product Number
Product Name
Product Description
Pricing
Two Natrix CH membrane chromatography devices: Natrix CH micro on the right and Natrix CH bench on the left
Natrix® CH chromatography membrane
Chromatographic purification is a frequent bottleneck in biomolecule purification. With high load capacity and ultra-fast flow rates, our Natrix® chromatography membranes deliver excellent impurity removal in a scalable, single-use format. This innovative technology solves many of your purification challenges and enables a new level of downstream productivity while improving process economics and facility utilization.
Two white vertical linear light bulbs against a plain white background. These fluorescent bulbs are designed with a long, slender tube and a base at the bottom for installation.
Natrix® Q chromatography membrane
A high-capacity, high-throughput, strong anion exchange membrane chromatography device designed for flow-through purification of biomolecules and removal of endotoxins, viruses, HCP and nucleic acids.
Natrix® Q chromatography membrane is a porous polyacrylamide hydrogel containing a high density of pendant quaternary ammonium (Q) binding groups directly polymerized within an inert, macroporous membrane scaffold. The interconnected pore structure and high functional group density enables fast flow rates with high loading capacity while maintaining excellent impurity removal across a wide range of buffer conditions. Natrix® Q chromatography membrane devices improve your productivity, flexibility, economic utility and process robustness.
Related Resources
Page 1 of 3