Skip to Content
Merck
All Photos(1)

Documents

RAD132

radiello Cartridge Adsorbents

for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

packaging

pk of 20

manufacturer/tradename

Radiello

technique(s)

passive air sampling: suitable

matrix

SS net with mix of mol sieve and activated charcoal (30-50 mesh)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling Anaesthetic Gases/Vapors

Looking for similar products? Visit Product Comparison Guide

General description

Several different Radiello cartridge adsorbents are available. Each of which are specific for sampling different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm D. Each cartridge is designed for one time use with the exception of Radiello thermal desorption (TD) cartridge adsorbents.

Each cartridge arrives in a sealed glass or plastic tube wrapped in a transparent thermally sealed polyethylene (PE) bag. The same sealed tube can be used to store the cartridge after sampling prior to desorption/analysis. A sufficient number of adhesive barcode labels are included with every pack of cartridges for easy tracking of sampling date and time.

Note: To conduct air sampling using the Radiello system, Radiello diffusive bodies and triangular support plates must be ordered (separately) in addition to cartridge adsorbents. Radiello Starter Kits (RADxxxS) include all the necessary parts to complete a sampling.

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Rocco Rella et al.
The Science of the total environment, 487, 255-259 (2014-05-03)
The proofing treatment of the floor slabs with a solvent based material and successive treatment with epoxy resin resulted in significant air contamination in a primary school. The contamination caused illnesses in many occupants of school. The GC/MS investigations of
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special

Articles

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Protocols

Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.

Related Content

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service