Skip to Content
Merck
All Photos(1)

Key Documents

59142

Supelco

SUPELCOSIL LC-ABZ (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
Pricing and availability is not currently available.

Product Name

SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

Agency

suitable for USP L60

Quality Level

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

12.0% carbon loading

parameter

≤70 °C temp. limit
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.4 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

amide, alkyl phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-ABZ phase has a unique deactivation technology which provides excellent reversed-phase performance for basic compounds, as well as those that are acidic, polar neutral, and non-polar. Silanol suppressing practices such as using competing amines, operating at high ionic strength, and adjusting mobile phases to pH extremes are typically not required to obtain good peak shape on the LC-ABZ phase.

Features and Benefits

• For acids, bases, zwitterions
• Increased polar retention relative to standard reversed-phases
• Unique selectivity
• Silanol deactivation
• Peak shape, efficiency, and retention are comparable to C8 phases for non-polar analytes

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Ufkes et al.
Pharmacy world & science : PWS, 20(2), 83-87 (1998-05-19)
To study the pharmacokinetics of dextromoramide in long-term opiate addicts on methadone maintenance therapy (MMT) a reverse-phase HPLC technique was developed to monitor dextromoramide and methadone concentrations in plasma simultaneously. After liquid-liquid extraction from plasma, dextromoramide and methadone were determined
Neil C Dias et al.
The Analyst, 128(5), 427-433 (2003-06-07)
The solvation parameter model is used to identify suitable chromatographic models for estimating the octanol-water partition coefficient for neutral compounds of varied structure by reversed-phase liquid chromatography. The stationary phase Supelcosil LC-ABZ with methanol-water mobile phases affords a series of
K Valkó et al.
Journal of pharmaceutical and biomedical analysis, 11(4-5), 361-366 (1993-04-01)
S-Adenosyl-L-methionine (SAM) and its metabolites S-adenosyl-L-homocysteine (SAH) and methyl-thioadenosine (MTA) are endogenous compounds that are heavily involved in a variety of biochemical processes, and have therefore been the target for several assays in body fluids and tissues. Reversed-phase chromatographic behaviour
D Korakas et al.
Journal of pharmaceutical and biomedical analysis, 12(9), 1141-1145 (1994-09-01)
A series of biologically active peptides and their conjugates with lipidic amino acids were investigated by systematic change of the mobile phase composition using traditional octadecylsilica stationary phase and the newly developed Supelcosil LC-ABZ column. The mobile phases contained various
J A López et al.
Journal of chromatography. A, 870(1-2), 105-114 (2000-03-18)
A high-performance liquid chromatographic method with electrochemical detection has been developed for the determination of six 1,4-dihydropyridines: nifedipine, nimodipine, nisoldipine, nicardipine, felodipine and lacidipine. The chromatographic separation was performed using a Supelcosil LC-ABZ+Plus C18 column. A mobile phase of methanol-water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service