Skip to Content
Merck
All Photos(1)

Key Documents

48179

Supelco

ASTM® D5307 Crude oil quantitative standard

6.25 % (w/w) each component, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D5307

CofA

current certificate can be downloaded

packaging

ampule of 2 mL

concentration

6.25 % (w/w) each component

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
ASTM® D5307 crude oil internal standard has been used in the quantitative determination of diamondoid compounds like adamantane in specific organic solvents like dichloromethane, hexane, carbon-tetrachloride, hexane , heptadecane, hexadecane, pentadecane and tetradecane, using infrared attenuated total reflection spectroscopy (IR-ATR).

Other Notes

Equal weights of the hydrocarbons listed. Use to determine detector response factors (slight warming of mixture may be necessary to dissolve all components completely.)

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Decane
    Dodecane
    Dotriacontane
    Eicosane
    Heptadecane
    Hexadecane
    Hexatriacontane
    Octacosane
    Octadecane
    Pentadecane
    Tetracontane
    Tetracosane
    Tetradecane
    Tetratetracontane
    Tridecane
    Undecane
    See All (16)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of adamantane in organic media via infrared attenuated total reflection spectroscopy
Luzinova Y, et al.
Organic Geochemistry, 40(11), 1143-1150 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service