Skip to Content
Merck
All Photos(1)

Key Documents

47864

Supelco

Nicotinic acid

analytical standard

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

feature

standard type water soluble vitamin

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

236-239 °C (lit.)

density

1.473 g/cm3 at 25 °C

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)
rat ... Chrnb2(54239)

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid acts as an antilipolytic drug in adipose tissue, and is commonly used for hyperlipoproteinaemia and pellagra.[1]
Nicotinic acid is a water soluble[2], vitamin belonging to B complex group of vitamins[3], which can be used as a broad spectrum lipid lowering drug.[4] Its mode of action basically involves the inhibition of the hormone-sensitive triglyceride lipase and thereby, decreasing the lipolysis in adipose tissue.[3][4]

Application

Nicotinic acid may be used as an analytical reference standard for the quantification of the analyte in tablet dosage forms using reversed-phase high-performance liquid chromatography technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nicotinic acid: the broad?spectrum lipid drug. A 50th anniversary review
Carlson.A.L
J. Int. Med., 258, 94-114 (2005)
PUMA-G and HM74 are receptors for nicotinic acid and mediate its anti-lipolytic effect
Tunaru S, et al.
Nature Medicine, 9, 352-352 (2003)
Molecular identification of nicotinic acid receptor
Soga T, et al.
Biochemical and Biophysical Research Communications, 303, 364-369 (2003)
RP-HPLC method for the determination of atorvastatin calcium and nicotinic acid in combined tablet dosage form
Shah DA, et al.
Indian Journal of Pharmaceutical Sciences, 69(5), 700-700 (2007)
John R Guyton et al.
Journal of the American College of Cardiology, 62(17), 1580-1584 (2013-08-07)
This study sought to examine the relationship between niacin treatment, lipoproteins, and cardiovascular (CV) outcomes in this secondary analysis of the AIM-HIGH (Atherothrombosis Intervention in Metabolic Syndrome With Low HDL/High Triglycerides and Impact on Global Health Outcomes) trial. During a

Questions

  1. questo standard è certificato iso17034 o iso 17025?

    1 answer
    1. Analytical standards are not ISO 17025 or 17034 certified. This level of certification is for a certified reference material such as product PHR1276.

      https://www.sigmaaldrich.com/product/sial/phr1276

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service