Skip to Content
Merck
All Photos(1)

Key Documents

20358

Supelco

ORBO 90 Carboxen® 564 (20/45), 160/80 mg

W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)

Synonym(s):

ORBO Carboxen-564 Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,F,F separators

Agency

NIOSH 2500

product line

ORBO

composition

Bed A, 160 mg
Bed B, 80 mg

packaging

pkg of 25 ea

manufacturer/tradename

ORBO 90

technique(s)

active air sampling: suitable

O.D. × L

6 mm × 75 mm

matrix

Carboxen® 564 Carbon Molecular Sieve (CMS)

particle size

20-45 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for analyte group MEK (methylethyl ketone)

Looking for similar products? Visit Product Comparison Guide

General description

  • ORBO 90 Carboxen® 564 (20/45), 160/80 mg solvent desorption tube is designed for gas and vapor sampling.
  • It contains two beds of the same selective adsorbent separated by glass wool or foam.
  • Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.

Legal Information

Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany
ORBO is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Leidy et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 26(4), 367-382 (1991-08-01)
A study was conducted to compare the efficiency of five adsorbents used by government and private laboratories to collect airborne pesticides. Six pesticides, acephate, chlordane, chlorpyrifos, diazinon, heptachlor, and propoxur, were vaporized in a closed system and collected on each
Sebastien Allard et al.
Journal of chromatography. A, 1238, 15-21 (2012-04-13)
Trihalomethanes are predominantly formed during disinfection of water via reactions of the oxidant with natural organic matter. Even though chlorinated and brominated trihalomethanes are the most widespread organic contaminants in drinking water, when iodide is present in raw water iodinated
Dawn M Chapman et al.
Journal of agricultural and food chemistry, 52(17), 5431-5435 (2004-08-19)
A rapid and automated solid phase microextraction (SPME) stable isotope dilution gas chromatography/mass spectrometry (GC-MS) method for 2-methoxy-3-isobutylpyrazine (MIBP) quantification in red wine was developed. Wines with 30% (w/v) NaCl and 2-methoxy-(2)H(3)-3-isobutylpyrazine internal standard were sampled with a 2 cm
Shih-Wei Tsai et al.
Journal of chromatography. A, 991(1), 1-11 (2003-04-22)
The solid-phase microextraction (SPME) device was used as a time-weighted average (TWA) sampler for ethylene oxide. Carboxen/polydimethylsiloxane (CAR/PDMS) fiber was used and hydrogen bromide (HBr) was loaded onto the fiber. The SPME fiber assembly was then inserted into PTFE tubing
Lucia Giordano et al.
Journal of chromatography. A, 1017(1-2), 141-149 (2003-10-31)
A new method was developed for the determination of 2-furfural (2-F) and 5-methylfurfural (5-MF), two products of Maillard reaction in vinegar, with head-space solid-phase microextraction (HS-SPME) coupled to gas chromatography-mass spectrometry (GC-MS). A divinylbenzene (DVB)/carboxen (CAR)/polydimethylsiloxane (PDMS) fibre was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service