Skip to Content
Merck
All Photos(3)

Key Documents

20231

Supelco

ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg

W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea

Synonym(s):

ORBO 2-HMP on Supelpak 20N Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W,W separators

Agency

EPA TO-15
OSHA 52,61

product line

Amberlite
ORBO

composition

Bed A, 150 mg
Bed B, 75 mg

packaging

pkg of 25 ea

manufacturer/tradename

ORBO 24

technique(s)

active air sampling: suitable

O.D. × L

6 mm × 105 mm

matrix

XAD-2 2-(Hydroxymethyl)piperidine (2-HMP) on XAD-2

particle size

20-40 mesh

application(s)

air monitoring
environmental
industrial hygiene

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
Supelpak is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Egea González et al.
Journal of AOAC International, 80(5), 1091-1097 (1997-11-05)
A method to sample and analyze chlorothalonil and dichlofluanid in greenhouse air was evaluated. Analysis was performed by gas chromatography with electron capture detection and gas chromatography-mass spectrometry. Solid sorbents such as Chromosorb 102, Porapak R, Supelpak-2, Amberlite XAD-2, Amberlite
Anne Ørbo et al.
The Journal of steroid biochemistry and molecular biology, 113(1-2), 139-149 (2009-01-14)
Patients with endometrial hyperplasia representing preliminary stages of endometrial cancer have shown to respond to therapy in 100% of the cases when treated with levonorgestrel-impregnated intrauterine device. Anti-proliferative effect has also been reported after application of an anti-progestin impregnated intrauterine
Anne Beate Vereide et al.
Gynecologic oncology, 97(3), 740-750 (2005-05-12)
The aim of the study was to investigate apoptosis as a growth regulatory mechanism of gestagen in endometrial precancers and to compare differences in the apoptotic cascade after high and low dose gestagen regimens. Pre- and post-treatment paraffin-embedded endometrial hyperplasia
H Stalsberg et al.
Virchows Archiv. A, Pathological anatomy and histopathology, 414(5), 415-422 (1989-01-01)
The autopsies of 12 victims from two snow avalanches in North-Norway are reported. Supportive evidence from non-autopsied and surviving victims is included. Consistent autopsy findings were prominent lung oedema, moderate cerebral oedema, extreme contraction of the left ventricle, petechiae in
K Flo et al.
Scandinavian journal of clinical and laboratory investigation, 55(8), 715-721 (1995-12-01)
Elevated extracellular cGMP levels have been observed in various clinical conditions, and the analyte has been proposed as a diagnostic marker of cardiovascular as well as malignant diseases. However, the use of extracellular cGMP as a pathophysiological marker requires detailed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service