Skip to Content
Merck
All Photos(1)

Key Documents

WH0003014M5

Sigma-Aldrich

Monoclonal Anti-H2AFX antibody produced in mouse

clone 3F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-H2A histone family, member X, Anti-H2A.X, Anti-H2A/X, Anti-H2AX

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... H2AFX(3014)

General description

H2AFX/H2AX (H2A histone family member X) codes for a key histone, that belongs to the histone H2A gene family. It is located on human chromosome 11q23.3.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif. (provided by RefSeq)

Immunogen

H2AFX (AAH04915, 1 a.a. ~ 143 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSGRGKTGGKARAKAKSRSSRAGLQFPVGRVHRLLRKGHYAERVGAGAPVYLAAVLEYLTAEILELAGNAARDNKKTRIIPRHLQLAIRNDEELNKLLGGVTIAQGGVLPNIQAVLLPKKTSATVGPKAPSGGKKATQASQEY

Application

Monoclonal Anti-H2AFX antibody has been used in western blotting.

Biochem/physiol Actions

H2AFX (H2A histone family member X) participates in the detection of DNA double-stranded breaks that can lead to translocations. Mutations in H2AFX gene results in non–hodgkin lymphoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Mahen et al.
PloS one, 8(5), e61893-e61893 (2013-05-10)
A-type lamins encoded by LMNA form a structural fibrillar meshwork within the mammalian nucleus. How this nuclear organization may influence the execution of biological processes involving DNA transactions remains unclear. Here, we characterize changes in the dynamics and biochemical interactions
Genetic variation in H2AFX contributes to risk of non-Hodgkin lymphoma
Novik KL, et al.
Cancer Epidemiology, Biomarkers & Prevention, 16(6), 1098-1106 (2007)
A-type lamins maintain the positional stability of DNA damage repair foci in mammalian nuclei
Mahen R, et al.
PLoS ONE (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service