Skip to Content
Merck
All Photos(4)

Key Documents

T1145

Sigma-Aldrich

L-Tyrosine disodium salt hydrate

BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)

Synonym(s):

L-3-(4-Hydroxyphenyl)alanine disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NNa2O3 · xH2O
CAS Number:
Molecular Weight:
225.15 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

color

white to beige

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].N[C@@H](Cc1ccc([O-])cc1)C([O-])=O

InChI

1S/C9H11NO3.2Na.H2O/c10-8(9(12)13)5-6-1-3-7(11)4-2-6;;;/h1-4,8,11H,5,10H2,(H,12,13);;;1H2/q;2*+1;/p-2/t8-;;;/m0.../s1

InChI key

SMJHYQCAVHUILN-CZDIJEQGSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tyrosine is a polar, non-essential amino acid with a phenolic functionality. L-tyrosine is a natural amino acid that is incorporated into proteins as directed by the genetic code. The phenolic function makes the tyrosine residue in proteins a principle recipient of reactions such as phosphorylation/dephosphorylation important for cell signalling. Tyrosine is also a precursor of various neurotransmitters and hormones.

Other Notes

Amino acid precursor of dopamine and other catecholamines

Caution

Extremely hygroscopic.

Suitability

This product has been qualified for use in insect cell cultures.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Wu et al.
Nature cell biology, 23(1), 75-86 (2021-01-10)
Nutrient availability is central for T-cell functions and immune responses. Here we report that CD8+ T-cell activation and anti-tumour responses are strongly potentiated by the non-essential amino acid Asn. Increased Asn levels enhance CD8+ T-cell activation and effector functions against
Ching-Hsia Hung et al.
European journal of pharmacology, 765, 457-462 (2015-09-17)
The purpose of the study was to estimate the ability of L-tyrosine to induce cutaneous analgesia and to investigate the interaction between L-tyrosine and the local anesthetic lidocaine. After subcutaneously injecting the rats with L-tyrosine and lidocaine in a dose-dependent
Zihao Wang et al.
Science signaling, 3(104), ra2-ra2 (2010-01-14)
Like phosphorylation, the addition of O-linked beta-N-acetylglucosamine (O-GlcNAcylation) is a ubiquitous, reversible process that modifies serine and threonine residues on nuclear and cytoplasmic proteins. Overexpression of the enzyme that adds O-GlcNAc to target proteins, O-GlcNAc transferase (OGT), perturbs cytokinesis and
Airi Sekine et al.
SpringerPlus, 4, 48-48 (2015-02-13)
The tryptophan metabolite, kynurenic acid (KYNA), is a preferential antagonist of the α7 nicotinic acetylcholine receptor at endogenous brain concentrations. Recent studies have suggested that increase of brain KYNA levels is involved in psychiatric disorders such as schizophrenia and depression.

Articles

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Learn more about the amino acid L-Tyrosine and explore how it is used in cell culture media like MEM, DMEM, and RPMI. We offer a comprehensive portfolio of amino acid cell culture supplements and reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service