Skip to Content
Merck
All Photos(1)

Key Documents

SRP3296

Sigma-Aldrich

WNT-7A human

recombinant, expressed in HEK 293 cells, ≥80% (SDS-PAGE), ≥80% (HPLC)

Synonym(s):

Human WNT-7A, Recombinant WNT-7A, WNT-7A Signaling Molecule

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥80% (HPLC)
≥80% (SDS-PAGE)

form

lyophilized

potency

40-60 ng/mL ED50

mol wt

35.5 kDa

packaging

pkg of 15 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WNT7A(7476)

General description

Wnt-7a is expressed in placenta, kidney, testis, uterus, fetal lung and fetal and adult brain. Most Wnt proteins can signal though a mechanism called the canonical Wnt pathway, in which Wnt proteins bind to and activate seven-pass transmembrane receptors of the Frizzled family ultimately leading to the disruption of β-Catenein degradation. Intracellular accumulation of β-Catenin increases translocation of the protein into the nucleus where it binds to TCF/LEF transcription factors and induces the expression of numerous genes. Increased Wnt/β-Catenein signaling is associated with tumorigenesis in a diverse set of human cancers. However, Wnt-7a/Frizzled-9 signaling has been shown to act as a tumor suppressor in non-small cell lung cancers.

Biochem/physiol Actions

Wnt-7a belongs to the Wnt family of signaling proteins that play a key role in maintaining the integrity of embryonic and adult tissues. Recombinant human Wnt-7a is a 35.5 kDa glycoprotein containing 318 amino acids. Due to glycosylation, Wnt-7a migrates between 40-55 kDa by SDS-PAGE gel under unreduced conditions

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of ≤0.2 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G T Wong et al.
Molecular and cellular biology, 14(9), 6278-6286 (1994-09-01)
The mouse Wnt family includes at least 10 genes that encode structurally related secreted glycoproteins. Wnt-1 and Wnt-3 were originally identified as oncogenes activated by the insertion of mouse mammary tumor virus in virus-induced mammary adenocarcinomas, although they are not

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service