Skip to Content
Merck
All Photos(1)

Documents

SRP0212

Sigma-Aldrich

PTP1B (1-321)/PTPN1 Active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

PTPN1, Protein tyrosine phosphatase, non-receptor type 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

63.3 kDa

packaging

pkg of 20 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.84 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPN1(5770)

General description

Protein tyrosine phosphatase, catalytic domain, (amino acids 1-321), (Gene bank accession no. M31724) with N-terminal GST tag, MW = 63.3 kDa, expressed in an E. coli expression system.
Protein tyrosine phosphatase, non-receptor type 1 (PTPN1), which is also known as protein-tyrosine phosphatase 1B (PTP1B), is a tyrosine phosphatase. It is localized at the cytoplasmic side of the endoplasmic reticulum. The gene encoding PTPN1 is localized on human chromosome 20q13.13.

Application

Useful for the study of enzyme kinetics, regulation, to dephosphorylate target substrates and for screening inhibitors.

Biochem/physiol Actions

Protein tyrosine phosphatase, non-receptor type 1 (PTPN1) or protein-tyrosine phosphatase 1B (PTP1B) dephosphorylates the epidermal growth factor receptor. It acts as a modulator of insulin-like growth factor receptor (IGFR) and MET-mediated signaling responses. It may be useful as a drug target in type 2 diabetes and obesity. The protein has been shown to enhance tumor progression in various cancers such as breast, colon and prostate. It acts a tumor suppressor in other cancers (lymphomas and esophageal cancers).

Unit Definition

One unit will hydrolyze 1 nmol p-nitrophenyl phosphate per minute at pH 7.4 and 30°C.

Physical form

Formulated in 25 Mm Tris-HCl, pH 8.0, 75 mM NaCl, 0.05% Tween-20, 50% glycerol, 2 mM EDTA, 1 mM DTT, and 10 mM glutathione.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

PTP1B-dependent regulation of receptor tyrosine kinase signaling by the actin-binding protein Mena.
Hughes SK
Molecular Biology of the Cell, 26, 3867-3878 (2015)
Mohammad Nasir Uddin et al.
Phytochemistry, 103, 99-106 (2014-05-13)
Protein tyrosine phosphatase 1B (PTP1B) plays a key role in metabolic signaling, thereby making it an exciting drug target for type 2 diabetes and obesity. Besides, there is substantial evidence that shows its overexpression is involved in breast cancer, which
Analysis of common PTPN1 gene variants in type 2 diabetes, obesity and associated phenotypes in the French population.
Cheyssac C
BMC Medical Genetics (2006)
Frequent amplification of PTP1B is associated with poor survival of gastric cancer patients.
Wang N
Cell Cycle, 14, 732-743 (2015)
Protein tyrosine phosphatase 1B (PTP1B): A key regulator and therapeutic target in liver diseases.
Chen PJ
Toxicology, 337, 10-20 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service