Skip to Content
Merck
All Photos(1)

Documents

SML2744

Sigma-Aldrich

Sitafloxacin sesquihydrate

≥98% (HPLC)

Synonym(s):

7-[(7S)-7-Amino-5-azaspiro[2.4]hept-5-yl]-8-chloro-6-fluoro-1-[(1R,2S)-2-fluorocyclopropyl]-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid, hydrate (2:3), DU 6859 sesquihydrate, DU 6859a sesquihydrate, DU-6859 sesquihydrate, DU-6859a sesquihydrate, DU6859 sesquihydrate, DU6859a sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18ClF2N3O3·1.5H2O
CAS Number:
Molecular Weight:
436.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

InChI

1S/C19H18ClF2N3O3.H2O/c20-14-15-8(17(26)9(18(27)28)5-25(15)12-4-10(12)21)3-11(22)16(14)24-6-13(23)19(7-24)1-2-19;/h3,5,10,12-13H,1-2,4,6-7,23H2,(H,27,28);1H2/t10-,12+,13+;/m0./s1

InChI key

ANCJYRJLOUSQBW-JJZGMWGRSA-N

Biochem/physiol Actions

Sitafloxacin (DU-6859 or DU-6859a) is an orally active, fourth generation fluoroquinolone that exhibits broad-spectrum antibiotic activity against many Gram-positive, Gram-negative and anaerobic clinical isolates, including strains resistant to other fluoroquinolones, via dual inhibitory activity against bacterial DNA gyrase and topoisomerase IV (IC50 = 1.16 and 1.88 μg/mL against S. pneumoniae gyrA/B and parC/E, respectively). Sitafloxacin is active against methicillin-resistant staphylococci, Streptococcus pneumoniae and other streptococci with reduced susceptibility to levofloxacin and other quinolones and enterococci. Sitafloxacin also exhibits activity against clinical isolates of Klebsiella pneumoniae, Enterobacter cloacae, Pseudomonas aeruginosa as well as some activity against quinolone-resistant strains and Acinetobacter baumannii. Sitafloxacin in vitro activity against anaerobes is comparable to imipenem and metronidazole.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lina Yi et al.
Journal of medical microbiology, 66(6), 770-776 (2017-06-10)
Purpose. Sitafloxacin (SFX) is a new fluoroquinolone (FQ) that has shown a strong bactericidal effect against Mycobacterium tuberculosis (Mtb) in vitro. However, data on SFX efficacy against Mtb with gyrA/B mutations and its epidemiological cut-off (ECOFF) value remain limited. Therefore
Ryoichi Hamasuna et al.
Microbial drug resistance (Larchmont, N.Y.), 24(1), 30-34 (2017-06-06)
Emergence of antimicrobial resistance in Neisseria gonorrhoeae is a major public health concern globally, and new antimicrobials for treatment of gonorrhea are imperative. In this study, the in vitro activity of sitafloxacin, a fluoroquinolone mainly used for respiratory tract or
Y Onodera et al.
The Journal of antimicrobial chemotherapy, 44(4), 533-536 (1999-12-10)
The in-vitro inhibitory activities of sitafloxacin (DU-6859a) and other quinolones against Streptococcus pneumoniae DNA gyrase and topoisomerase IV were measured. IC50s of levofloxacin, ciprofloxacin, sparfloxacin and tosufloxacin against DNA gyrase were almost three to 12 times higher than those against
Xiaomeng Dong et al.
International journal of clinical and experimental medicine, 8(5), 8135-8140 (2015-07-30)
To detect the in vitro activities of sitafloxacin alone and in combination with rifampin, colistin, sulbactam, and tigecycline against extensively drug-resistant Acinetobacter baumannii (XDR-A. baumannii). 24 XDR-A. baumannii strains were isolated from patients' specimens. Broth microdilution assay was used to
K Sato et al.
Antimicrobial agents and chemotherapy, 36(7), 1491-1498 (1992-07-01)
DU-6859, (-)-7-[(7S)-amino-5-azaspiro(2,4)heptan-5-yl]-8-chloro-6- fluoro-1-[(1R,2R)-cis-2-fluoro-1-cyclopropyl]-1,4-dihydro-4-oxoquinol one-3- carboxylic acid, is a new fluoroquinolone with antibacterial activity which is significantly better than those of currently available quinolones. The MICs for 90% of methicillin-susceptible and -resistant Staphylococcus aureus and Staphylococcus epidermidis clinical isolates (MIC90s) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service