Skip to Content
Merck
All Photos(1)

Documents

SML1072

Sigma-Aldrich

GSK-LSD1

≥98% (HPLC)

Synonym(s):

GSK-LSD1 (trans-racemic) dihydrochloride, rel- N-[(1R,2S)-2-Phenylcyclopropyl]-4-Piperidinamine hydrochloride (1:2)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2 · 2HCl
CAS Number:
Molecular Weight:
289.24
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

C1(N[C@@H]2C[C@H]2C3=CC=CC=C3)CCNCC1

InChI

1S/C14H20N2/c1-2-4-11(5-3-1)13-10-14(13)16-12-6-8-15-9-7-12/h1-5,12-16H,6-10H2/t13-,14+/m0/s1

InChI key

BASFYRLYJAZPPL-UONOGXRCSA-N

Application

GSK-LSD1 has been used as a lysine specific demethylase 1 (LSD1) inhibitor in mice to study sepsis-induced mortality. It has also been used as an inhibitor to study if the effect of phenelzine activation of human immunodeficiency virus (HIV) latency is caused due to LSD1.

Biochem/physiol Actions

GSK-LSD1 is a potent and selective inhibitor of lysine specific demethylase 1 (LSD1). GSK-LSD1 potently inhibits proliferation of varies cancer cell lines by changing gene expression patterns. For full characterization details, please visit the GSK-LSD1 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

GSK-LSD1 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaeyoon Oh et al.
BMB reports, 53(7), 385-390 (2020-04-23)
Inflammatory Bowel Disease is caused by an acute or chronic dysfunction of the mucosal inflammatory system in the intestinal tract. In line with the results of our previous study, wherein we found that the PKCα-LSD1-NF-κB signaling plays a critical role
PKCα-LSD1-NF-κB-Signaling Cascade Is Crucial for Epigenetic Control of the Inflammatory Response
Kim D, et al.
Molecular Cell, 69(3), 398-411 (2018)
HIV-1 infection of microglial cells in a reconstituted humanized mouse model and identification of compounds that selectively reverse HIV latency
Llewellyn GN, et al.
Journal of Neurovirology, 1-12 (2017)
Carl M Gay et al.
Cancer cell, 39(3), 346-360 (2021-01-23)
Despite molecular and clinical heterogeneity, small cell lung cancer (SCLC) is treated as a single entity with predictably poor results. Using tumor expression data and non-negative matrix factorization, we identify four SCLC subtypes defined largely by differential expression of transcription
Cavan P Bailey et al.
Frontiers in immunology, 11, 2196-2196 (2020-10-13)
Cell therapies such as chimeric-antigen receptor (CAR) T-cells and NK cells are cutting-edge methods for treating cancer and other diseases. There is high interest in optimizing drug treatment regimens to best work together with emerging cell therapies, such as targeting

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service