Skip to Content
Merck
All Photos(1)

Key Documents

SML1042

Sigma-Aldrich

Eptifibatide acetate

≥98% (HPLC)

Synonym(s):

Cyclo(S,S)-Mpr(Har)GDWPC-NH2 acetate, N6-(Aminoiminomethyl)-N2-(3-mercapto-1-oxopropyl)-L-lysylglycyl-L-α-aspartyl-L-tryptophyl-L-prolyl-L-cysteinamide cyclic (16)-disulfide acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H49N11O9S2 · xC2H4O2
CAS Number:
Molecular Weight:
831.96 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

General description

Eptifibatide acetate is a cyclic heptapeptide. It is the member of the class arginin-glycin-aspartat-mimetics. Eptifibatide has a short half-life. The drug decreases the risk of myocardial infarction (MI). Eptifibatide acetate might be associated with thrombocytopenia.

Biochem/physiol Actions

Eptifibatide is a specific inhibitor of fibrinogen binding to the platelet membrane integrin glycoprotein GPIIb/IIIa receptor, preventing platelet aggregation. Eptifibatide binds reversibly and competitively.
Eptifibatide is a specific inhibitor of fibrinogen binding to the platelet membrane integrin glycoprotein GPIIb/IIIa receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Recurrent acute thrombocytopenia in the hospitalized patient: Sepsis, DIC, HIT, or antibiotic-induced thrombocytopenia
Rousan TA, et al.
American Journal of Hematology, 85(1), 71-74 (2010)
Clinical review: traumatic brain injury in patients receiving antiplatelet medication
Beynon C, et al.
Critical Care, 16(4), 228-228 (2012)
Current role of platelet glycoprotein IIb/IIIa inhibitors in acute coronary syndromes
Bhatt DL and Topol EJ
JAMA : The Journal of the American Medical Association, 284(12), 1549-1558 (2000)
Eptifibatide: A cyclic peptide that selectively inhibits platelet glycoprotein IIb/IIIa
Heras M and Escolar G
Drugs Today (Barc), 36, 295-311 (2000)
Jaqueline Gomes Rosa et al.
Toxicon : official journal of the International Society on Toxinology, 159, 50-60 (2019-01-25)
Victims of Bothrops jararaca snakebites manifest bleedings, blood incoagulability, platelet dysfunction, and thrombocytopenia, and the latter has been directly implicated in the genesis of hemorrhagic diathesis. We addressed herein the direct effects of B. jararaca venom (BjV) on ex vivo

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service