Skip to Content
Merck
All Photos(1)

Key Documents

SML0684

Sigma-Aldrich

SF1670

≥98% (HPLC)

Synonym(s):

N-(9,10-Dihydro-9,10-dioxo-2-phenanthrenyl)-2,2-dimethyl-propanamide, N-(9,10-Dioxo-9,10-dihydro-phenanthren-2-yl)-2,2-dimethylpropionamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO3
CAS Number:
Molecular Weight:
307.34
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, light pink to red to very dark purple

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c1cc2c(cc1)c3c(cccc3)C(=O)C2=O)C(=O)C(C)(C)C

InChI

1S/C19H17NO3/c1-19(2,3)18(23)20-11-8-9-13-12-6-4-5-7-14(12)16(21)17(22)15(13)10-11/h4-10H,1-3H3,(H,20,23)

InChI key

VZQDDSYKVYARDW-UHFFFAOYSA-N

Application

SF1670 has been used:
  • To study the effect of microRNA-30a on PTEN (phosphatase and tensin homologue deleted on chromosome 10 ) expression regulation.
  • To study the formation of autophagosomes, induced by EGF (Epidermal growth factor) - dependent PTEN expression.
  • To study the role of PTEN in the regulation of autophagy induction in human normal endometrial stromal cells.

Biochem/physiol Actions

PTEN prevents PI3-K(phosphoinositide 3-kinase)/Akt signaling pathway, which is very essential for neuronal regeneration. Therefore, inhibition of PTEN by SF1670 may support neuronal regeneration.
SF1670 is a selective PTEN (phosphatase and tensin homolog) inhibitor with an IC50 of 2 micromolar. PTEN inhibition by SF1670 was shown to increase neutrophil activity (polarization, phagocytosis, oxidative burst, and recruitment to site of infection), thereby enhancing the inflammatory response and bacteria-killing capacity of neutopenic mice.
SF1670 is a selective PTEN (phosphatase and tensin homolog) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bioinformatics prediction of miR-30a targets and its inhibition of cell proliferation of osteosarcoma by up-regulating the expression of PTEN.
Zhong B, et al.
BMC Medical Genomics, 10(1), 64-64 (2017)
PTEN inhibition to facilitate intrinsic regenerative outgrowth of adult peripheral axons.
Christie K J, et al.
The Journal of Neuroscience, 30(27), 9306-9315 (2010)
Paradoxical induction of growth arrest and apoptosis by EGF via the up-regulation of PTEN by activating Redox factor-1/Egr-1 in human lung cancer cells.
Ryu J W, et al.
Oncotarget, 8(3), 4181-4181 (2017)
Aberrant PTEN expression in response to progesterone reduces endometriotic stromal cell apoptosis.
Choi J, et al.
Reproduction (Cambridge, England), 153(1), 11-21 (2017)
Stefanie Kälin et al.
Cell metabolism, 26(3), 475-492 (2017-09-07)
Obesity and type 2 diabetes are associated with metabolic defects and adipose tissue inflammation. Foxp3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service