Skip to Content
Merck
All Photos(1)

Key Documents

SMB00224

Sigma-Aldrich

3,4-Di-O-caffeoylquinic acid

≥90% (LC/MS-ELSD)

Synonym(s):

3,4-DICQA, 3,4-Di-O-caffeoylquinate, 3,4-Dicaffeoylquinic acid, Isochlorogenate B, Isochlorogenic acid B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1OC(=O)\C=C\c3ccc(O)c(O)c3)C(O)=O

InChI

1S/C25H24O12/c26-15-5-1-13(9-17(15)28)3-7-21(31)36-20-12-25(35,24(33)34)11-19(30)23(20)37-22(32)8-4-14-2-6-16(27)18(29)10-14/h1-10,19-20,23,26-30,35H,11-12H2,(H,33,34)/b7-3+,8-4+/t19-,20-,23-,25+/m1/s1

InChI key

UFCLZKMFXSILNL-PSEXTPKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Di-O-caffeoylquinic acid (3,4-COQ) is a component of the medicinal herb Youngia japonica.It has a caffeoyl moiety attached with a hexacyclic quinic acid in its structure.

Application

3,4-Di-O-caffeoylquinic acid has been used as a reference standard:
  • to quantify the caffeoylquinic acids of Artemisia frigida Willd. (Fringed sagewort) using high-performance liquid chromatography with diode array detection and electrospray ionization triple quadrupole mass spectrometric detection (HPLC-DAD-ESI-QQQ-MS)
  • to quantify the phenolic compounds of Artemisia species using high-performance liquid chromatography with diode array detection (HPLC-DAD) technique
  • for metabolic profiling of methanolic leaf extract of A. nallamalayana and to identify the phenolic acids using High-Performance Liquid Chromatography-Ultraviolet (HPLC-UV) analysis

Biochem/physiol Actions

3,4-Di-O-caffeoylquinic acid, along with 3,5-dicaffeoylquinic acid and luteolin-7-O-glucoside, is a component of the medicinal herb Youngia japonica. It has known antiviral activity and along with 3,5,-dicaffeoylquinic acid, shows activity against respiratory syncyntial virus (RSV). 3,4-COQ elicits antioxidant effects which mediate its neuroprotective effects against in vitro retinal damage. It also exerts cytoprotective effects against oxidative damage in bone marrow-derived mesenchymal stem cells (bmMSCs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antiviral activity and mode of action of caffeoylquinic acids from Schefflera heptaphylla (L.) Frodin.
Li Y, et al.
Antiviral Research, 68(1), 1-9 (2005)
Esterification of the Free Carboxylic Group in 3,4-di-O-caffeoylquinic Acid Enhances the Inhibition Activity against Respiratory Syncytial Virus (RSV)
Wu D, et al.
Medicinal Chemistry (2015)
A comparative assessment of in vitro cytotoxic activity and phytochemical profiling of Andrographis nallamalayana JL Ellis and Andrographis paniculata (Burm. f.) Nees using UPLC-QTOF-MS/MS approach
Goel N, et al.,
Royal Society of Chemistry Advances, 11(57), 35918-35936 (2021)
Yoshimi Nakajima et al.
Life sciences, 80(4), 370-377 (2006-10-19)
We investigated whether water extract of Brazilian green propolis (WEP) and its main constituents [caffeoylquinic acid derivatives (3,4-di-O-caffeoylquinic acid, 3,5-di-O-caffeoylquinic acid, chlorogenic acid) and cinnamic acid derivatives (p-coumaric acid, artepillin C, drupanin, baccharin)] exert neuroprotective effects against the retinal damage
Xican Li et al.
Molecules (Basel, Switzerland), 23(1) (2018-01-25)
In this study, a series of di-O-caffeoylquinic acids (di-COQs) were systematically investigated for their antioxidant and cytoprotective effects towards •OH-damaged bone marrow-derived mesenchymal stem cells (bmMSCs). Five di-COQs were measured using a set of antioxidant assays. The results show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service