Skip to Content
Merck
All Photos(1)

Key Documents

SAE0088

Sigma-Aldrich

Filipin complex ready made solution

from Streptomyces filipinensis, 5mg/ml in DMSO based solution

Synonym(s):

Filipin solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥90% (HPLC)

form

ready-to-use solution

storage condition

protect from light

shipped in

dry ice

storage temp.

−20°C

General description

Filipin Complex, the collective name given to family of isomeric polyene macrolides isolated from cultures of S. filipinensis, with Filipin III being the majority part of the complex.

Application

Filipin complex readymade solution has been used in the staining of HeLa cells for cytoskeletal imaging.
Filipin has been used in a double staining procedure as a probe for the detection of lipoproteins in polyacrylamide gel and immobilized on nitrocellulose membranes. It is also widely used to localize and quantitate unesterified cholesterol by virtue of a specific fluorescent complex.

Biochem/physiol Actions

Filipin is an antibiotic with antifungal functionality. Filipin complex has been found to alter cell membrane structure by interacting with the membrane sterols ergosterols and cholesterol. On binding to cholesterol, filipin shows fluorescence shift. As such, filipin complex has become a useful tool for diagnosis of Niemann-Pick type C disease, and for detection and quantification of cholesterol in cell membranes and lipid quantification in brain.

Other Notes

Filipin complex ready made solution is very sensitive to air and light. Upon receipt aliquot and store at -20°C. Avoid freeze thaw cycles.
Filipin complex is used in various concentrations depending on the specific protocol used. Dilute 5mg/ml stock solution with appropriate buffer according to the used protocol.
Filipin complex interaction with cholesterol alters the absorption and fluorescence spectra, for visualization with a fluorescence microscope use excitation at 340-380 nm and emission at 385-470 nm (Filipin fluorescent staining photo-bleaches very rapidly, thus the sample should be analyzed immediately).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi Zhao et al.
Redox biology, 62, 102678-102678 (2023-03-21)
Elevated lipid peroxidation (LPO), usually present in the tumour microenvironment (TME), is profoundly implicated in antitumour immunity and may be targeted for the development of new antitumour therapies. However, tumour cells may also rewire their metabolism to survive elevated LPO.
The pentaene macrolide antibiotic filipin prefers more rigid DPPC bilayers: a fluorescence pressure dependence study
Castanho MRB, et al.
Biochimica et Biophysica Acta - Biomembranes, 1419(1), 1-14 (1999)
Zoe White et al.
Skeletal muscle, 12(1), 25-25 (2022-11-30)
Limb-girdle muscular dystrophy (MD) type 2B (LGMD2B) and Duchenne MD (DMD) are caused by mutations to the Dysferlin and Dystrophin genes, respectively. We have recently demonstrated in typically mild dysferlin- and dystrophin-deficient mouse models that increased plasma cholesterol levels severely
The filipin complex.
M E Bergy et al.
Biochemistry, 7(2), 653-659 (1968-02-01)
Hailan He et al.
CNS neuroscience & therapeutics, 29(12), 4059-4069 (2023-06-29)
Christianson syndrome (CS) is caused by mutations in SLC9A6 and is characterized by global developmental delay, epilepsy, hyperkinesis, ataxia, microcephaly, and behavioral disorder. However, the molecular mechanism by which these SLC9A6 mutations cause CS in humans is not entirely understood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service