SAB4600388
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF™488A antibody produced in goat
~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Recommended Products
biological source
goat
Quality Level
conjugate
CF™ 488A conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
mouse
concentration
~2 mg/mL
technique(s)
flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 1-10 μg/mL
western blot: suitable
fluorescence
λex 490 nm; λem 515 nm
storage temp.
−20°C
target post-translational modification
unmodified
Related Categories
General description
Digestion of IgG by papain results in generation of fragment antigen binding (Fab), comprising of one complete L chain and a variable and CH1 region of H chain. Pepsin digestion of IgG results in fragment crystallizable (fc), comprising of the H chain constant region. IgG1 is the most abundant antibody and its deficiency results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions and IgG4 is associated with asymptomatic infection. IgG antibody has enormous therapeutic potential and the Fc region is used for the development of therapeutic antibodies.
This product is prepared by labeling high quality F(ab′)2 fragment of goat anti-mouse IgG (H+L) with the green fluorescent dye CF488A. CF488A is a superior alternative to the traditional green fluorescent dye FITC (or fluorescein) owing to its exceptional brightness and photostability. CF488A is comparably bright and photostable to Alexa Fluor 488. However, antibody conjugates prepared from CF488A are more specific than those prepared from Alexa Fluor 488 because CF488A, being minimally charged, does not significantly change the isoelectric points of antibodies as Alexa Fluor 488 does.
Specificity
Binds with whole molecule mouse IgG and the light chains of other mouse immunoglobulins.
Immunogen
mouse IgG(H+L)
Application
Anti-Mouse IgG (H+L), F(ab?)2 fragment, CF™488A antibody produced in goat has been used in the detection of CF488 in human cell lines using confocal fluorescence microscopy and in vinculin cytoskeleton using confocal immunofluorescence imaging.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 0.2% BSA.
Preparation Note
Protect from light. The antibody solution should be gently mixed before use.
Legal Information
This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
CF is a trademark of Biotium, Inc.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
E-cadherin-mediated force transduction signals regulate global cell mechanics
Muhamed I, et al.
Journal of Cell Science, jcs-185447 (2016)
AuapNIPAM SERRS tags for multiplex immunophenotyping cellular receptors and imaging tumor cells
Bodelon G, et al.
Small, 11(33), 4149-4157 (2015)
Stig Skrivergaard et al.
International journal of molecular sciences, 22(16) (2021-08-28)
Cultured meat is an emerging alternative food technology which aims to deliver a more ethical, sustainable, and healthy muscle-tissue-derived food item compared to conventional meat. As start-up companies are rapidly forming and accelerating this technology, many aspects of this multi-faceted
Sergio Rodal-Cedeira et al.
ACS nano, 14(11), 14655-14664 (2020-09-02)
Raman-encoded gold nanoparticles (NPs) have been widely employed as photostable multifunctional probes for sensing, bioimaging, multiplex diagnostics, and surface-enhanced Raman scattering (SERS)-guided tumor therapy. We report a strategy toward obtaining a particularly large library of Au nanocapsules encoded with Raman
Jennifer Batalla-Covello et al.
Cancers, 13(24) (2021-12-25)
Tumor tropic neural stem cells (NSCs) can improve the anti-tumor efficacy of oncovirotherapy agents by protecting them from rapid clearance by the immune system and delivering them to multiple distant tumor sites. We recently completed a first-in-human trial assessing the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service