Skip to Content
Merck
All Photos(1)

Documents

SAB4501891

Sigma-Aldrich

Anti-MMP-2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

72 kDa gelatinase, 72 kDa type IV collagenase, CLG4, CLG4A, collagenase type IV-A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 73 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP2(4313)

General description

Anti-MMP-2 Antibody detects endogenous levels of total MMP-2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human MMP-2.

Immunogen Range: 611-660

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fen-Fen Liang et al.
PloS one, 8(10), e76517-e76517 (2013-10-12)
After years of experiments and clinical studies, parathyroid hormone-related protein(PTHrP) has been shown to be a bone formation promoter that elicits rapid effects with limited adverse reaction. Recently, PTHrP was reported to promote fibrosis in rat kidney in conjunction with
Fan Deng et al.
Annals of translational medicine, 10(8), 472-472 (2022-05-17)
Airway inflammation produced by neutrophils is a critical factor in the development of chronic obstructive pulmonary disease (COPD). Poor or excessive neutrophil polarization and chemotaxis may lead to pathogen accumulation and tissue damage. However, it is unclear how cigarette smoke
Pengyu Liu et al.
OncoTargets and therapy, 13, 4099-4110 (2020-06-05)
This study aimed to investigate the regulatory role and mechanism of microRNA-766 (miR-766) on cutaneous squamous cell carcinoma (CSCC) cells. The expression of miR-766 and programmed cell death 5 (PDCD5) was detected in CSCC tissues and CSCC cell lines (A431
Qiang Zhang et al.
Chinese medical journal, 135(9), 1076-1086 (2022-02-23)
Fibrosis in the peripheral airways contributes to airflow limitation in patients with chronic obstructive pulmonary disease (COPD). However, the key proteins involved in its development are still poorly understood. Thus, we aimed to identify the differentially expressed proteins (DEPs) between
Huaming Wei et al.
OncoTargets and therapy, 13, 13401-13411 (2021-01-08)
To investigate the effect of TRIM47 on glioma cells and further explore its underlying molecular mechanisms. Mouse xenograft model was used in this study. The mRNA expression of TRIM47 was detected by qRT-PCR. The cell viability and proliferation activity was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service