Skip to Content
Merck
All Photos(4)

Documents

SAB1412356

Sigma-Aldrich

ANTI-PAX7 antibody produced in mouse

clone 1G11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

HUP1, PAX7, PAX7B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G11, monoclonal

form

buffered aqueous solution

mol wt

antigen 37.84 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAX7(5081)

General description

Paired box 7 (PAX7) is encoded by the gene mapped to human chromosome 1p36.13. The encoded protein belongs to the PAX family and is expressed in central nervous system (CNS), craniofacial tissue, somites/skeletal muscle. PAX7 protein consists of paired domain (PD), an octapeptide (OP) motif and the helix-turn-helix motif of the homeodomain 1, 2 and 3 (HD1/2/3).
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

PAX7 (NP_002575.1, 411 a.a. ~ 520 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GGLDSATSISASCSQRADSIKPGDSLPTSQAYCPPTYSTTGYSVDPVAGYQYGQYGQSECLVPWASPVPIPSPTPRASCLFMESYKVVSGWGMSISQMEKLKSSQMEQFT

Biochem/physiol Actions

Paired box 7 (PAX7) in muscle-derived stem cells, plays an essential role in myogenic satellite cell specification, by inhibiting alternate developmental programs. Variation in the gene expression is associated with melanoma, neuroblastoma, rhabdomyosarcoma. PAX7 and PAX3 is also implicated in skeletal muscle formation and regeneration.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret Buckingham et al.
Annual review of cell and developmental biology, 23, 645-673 (2007-05-18)
Pax genes play key roles in the formation of tissues and organs during embryogenesis. Pax3 and Pax7 mark myogenic progenitor cells and regulate their behavior and their entry into the program of skeletal muscle differentiation. Recent results have underlined the
T H Beaty et al.
Human genetics, 132(7), 771-781 (2013-03-21)
A collection of 1,108 case-parent trios ascertained through an isolated, nonsyndromic cleft lip with or without cleft palate (CL/P) was used to replicate the findings from a genome-wide association study (GWAS) conducted by Beaty et al. (Nat Genet 42:525-529, 2010)
P Seale et al.
Cell, 102(6), 777-786 (2000-10-13)
The paired box transcription factor Pax7 was isolated by representational difference analysis as a gene specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization revealed that Pax7 was also expressed in satellite cells residing in adult muscle. Cell culture
Cecilia Romagnoli et al.
International journal of molecular sciences, 22(14) (2021-07-25)
Skeletal muscle has an outstanding capacity for regeneration in response to injuries, but there are disorders in which this process is seriously impaired, such as sarcopenia. Pharmacological treatments to restore muscle trophism are not available, therefore, the identification of suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service