Skip to Content
Merck
All Photos(2)

Key Documents

SAB1411338

Sigma-Aldrich

Anti-TDO2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

TDO, TPH2, TRPO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 47.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TDO2(6999)

General description

Tryptophan 2,3-dioxygenase (EC 1.13.11.11) plays a role in catalyzing the first and rat-limiting step in the kynurenine pathway, the major pathway of tryptophan metabolism.[supplied by OMIM

Immunogen

TDO2 (NP_005642.1, 1 a.a. ~ 406 a.a) full-length human protein.

Sequence
MSGCPFLGNNFGYTFKKLPVEGSEEDKSQTGVNRASKGGLIYGNYLHLEKVLNAQELQSETKGNKIHDEHLFIITHQAYELWFKQILWELDSVREIFQNGHVRDERNMLKVVSRMHRVSVILKLLVQQFSILETMTALDFNDFREYLSPASGFQSLQFRLLENKIGVLQNMRVPYNRRHYRDNFKGEENELLLKSEQEKTLLELVEAWLERTPGLEPHGFNFWGKLEKNITRGLEEEFIRIQAKEESEEKEEQVAEFQKQKEVLLSLFDEKRHEHLLSKGERRLSYRALQGALMIYFYREEPRFQVPFQLLTSLMDIDSLMTKWRYNHVCMVHRMLGSKAGTGGSSGYHYLRSTVSDRYKVFVDLFNLSTYLIPRHWIPKMNPTIHKFLYTAEYCDSSYFSSDESD

Biochem/physiol Actions

TDO2 (tryptophan 2, 3-dioxygenase) is responsible for the cleavage of indole ring at the C2-C3 bond of L-tryptophan, thereby regulating tryptophan concentration. It is expressed in tumor cells and suppresses antitumor immune responses. During acute ethanol intake, TDO2 carries the kynurenine pathway via glucocorticoid-associated route. This can lead to the accumulation of neurotoxic metabolites along with the reduction of serotonin. The TDO2 gene is upregulated in brains of patients with Alzheimer′s disease and might be associated with making of neurofibrillary tangles and senile plaque. TDO2 is also responsible for antimicrobial and immunoregulatory activities.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia K Schmidt et al.
European journal of immunology, 39(10), 2755-2764 (2009-07-29)
In mammals, the regulation of local tryptophan concentrations by the IFN-gamma-i inducible enzyme IDO is a prominent antimicrobial and immunoregulatory effector mechanism. Here, we show for the first time that another tryptophan-degrading enzyme, the liver-specific tryptophan 2,3-dioxygenase (TDO), is also
Bing Meng et al.
Proteins, 82(11), 3210-3216 (2014-07-30)
Tryptophan 2,3-dioxygenase (TDO), one of the two key enzymes in the kynurenine pathway, catalyzes the indole ring cleavage at the C2-C3 bond of L-tryptophan. This is a rate-limiting step in the regulation of tryptophan concentration in vivo, and is thus
Marion Soichot et al.
Alcohol and alcoholism (Oxford, Oxfordshire), 48(4), 415-425 (2013-04-06)
In response to acute ethanol consumption, tryptophan 2,3-dioxygenase (TDO) induces the kynurenine pathway (KP) through a glucocorticoid-mediated mechanism, which could lead to a dramatic accumulation of neurotoxic metabolites in association with serotonin depletion. As a result, interindividual variability in ethanol-induced
Olga Novikov et al.
Molecular pharmacology, 90(5), 674-688 (2016-10-19)
The endogenous ligand-activated aryl hydrocarbon receptor (AHR) plays an important role in numerous biologic processes. As the known number of AHR-mediated processes grows, so too does the importance of determining what endogenous AHR ligands are produced, how their production is
Wei Wu et al.
PloS one, 8(4), e59749-e59749 (2013-05-01)
To assess the role of the kynurenine pathway in the pathology of Alzheimer's disease (AD), the expression and localization of key components of the kynurenine pathway including the key regulatory enzyme tryptophan 2,3 dioxygenase (TDO), and the metabolites tryptophan, kynurenine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service