Skip to Content
Merck
All Photos(1)

Documents

SA20

Supelco

Starch (HK) Assay Kit

sufficient for 20 assays

Synonym(s):

Starch Quantitation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.84

usage

sufficient for 20 assays

analyte chemical class(es)

sugars

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Application

This kit is for the quantitative, enzymatic determination of native starch in food and other materials using the amyloglucosidase-hexokinase-glucose 6-phosphate dehydrogenase enzymatic assay. The hydrolysis of starch to glucose is catalyzed by amyloglucosidase. Glucose is phosphorylated by adenosine triphosphate (ATP) in the reaction catalyzed by hexokinase. Glucose-6-phosphate (G6P) is then oxidized to 6-phosphogluconate in the presence of nicotinamide adenine dinucleotide (NAD) in a reaction catalyzed by glucose-6-phosphate dehydrogenase (G6PDH). During this oxidation, an equimolar amount of NAD is reduced to NADH. The increase in absorbance at 340 nm is directly proportional to the glucose concentration.

Kit Components Only

Product No.
Description

  • glucose assay reagent 20 mL

  • starch assay reagent 1 mL/vial

  • starch assay standard 5 g

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taotao Zhu et al.
Molecular plant, 13(11), 1624-1643 (2020-09-22)
Genic male sterility (GMS) is critical for heterosis utilization and hybrid seed production. Although GMS mutants and genes have been studied extensively in plants, it has remained unclear whether chloroplast-associated photosynthetic and metabolic activities are involved in the regulation of
Zhongqi He et al.
Molecules (Basel, Switzerland), 27(1) (2022-01-12)
Common "glanded" (Gd) cottonseeds contain the toxic compound gossypol that restricts human consumption of the derived products. The "glandless" (Gl) cottonseeds of a new cotton variety, in contrast, show a trace gossypol content, indicating the great potential of cottonseed for
B Degrenne et al.
Biotechnology and bioengineering, 108(10), 2288-2299 (2011-04-27)
Photosynthetic hydrogen production under light by the green microalga Chlamydomonas reinhardtii was investigated in a torus-shaped PBR in sulfur-deprived conditions. Culture conditions, represented by the dry biomass concentration of the inoculum, sulfate concentration, and incident photon flux density (PFD), were
M Djanaguiraman et al.
Plant, cell & environment, 41(5), 993-1007 (2017-02-09)
The objectives were to (1) quantify high temperature (HT) stress impacts at different growth stages (season long, booting to seed-set and booting to maturity) on various yield components; (2) identify the most sensitive stage(s) to short episodes of HT stress
Amelia Torcello-Gómez et al.
Food chemistry, 272, 18-25 (2018-10-13)
An innovative procedure for plant chloroplasts isolation has been proposed, which consists of juice extraction by physical fractionation from plant material and recovery of its chloroplast-rich fraction (CRF) by centrifugation. This simple method has been applied to pea vine haulm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service