Skip to Content
Merck
All Photos(1)

Key Documents

S6016

Sigma-Aldrich

Wheaton Coplin staining jars

8 slide jar with glass cover

Synonym(s):

900620

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
MA.01

material

glass jar

feature

capacity 8 3 × 1 in. (76 × 25 mm) slide (sixteen slides back-to-back)
Wheaton staining ware is not recommended for any moist heat applications such as microwaves, autoclaves or water baths.

packaging

pack of 6 ea

manufacturer/tradename

WHEATON® 900620

dish L × W

47 mm × 26 mm

dish capacity

3 × 1 in. (76 × 25 mm) , 8 single slides or 16 slides back-to-back

inside L × W × D

42 mm × 26 mm × 85 mm

Looking for similar products? Visit Product Comparison Guide

General description

The wide top makes slide transfer easy and is suitable for staining slides that are inscribed on one end.

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.

Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.

Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.

Available Fluorescent in situ hybridization (FISH) procedures, reagents and equipment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service