Skip to Content
Merck
All Photos(1)

Key Documents

RAB1170

Sigma-Aldrich

Human PLG (Plasminogen) ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum sample(s)
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.14 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PLG(5340)

General description

This ELISA antibody pair detects Human Plasminogen (PLG)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masafumi Fukagawa et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 32(10), 1723-1730 (2017-01-07)
Secondary hyperparathyroidism (SHPT) is a major complication associated with chronic kidney disease. We evaluated the efficacy and safety of etelcalcetide (ONO-5163/AMG 416), a novel intravenous calcimimetic, in Japanese haemodialysis patients with SHPT. In this phase 3, multicentre, randomized, double-blind, placebo-controlled
Kelly M Shepardson et al.
Frontiers in immunology, 9, 2589-2589 (2018-11-27)
Influenza virus infections particularly when followed by bacterial superinfections (BSI) result in significant morbidities and mortalities especially during influenza pandemics. Type I interferons (IFNs) regulate both anti-influenza immunity and host susceptibility to subsequent BSIs. These type I IFNs consisting of
Terhi Ruohtula et al.
Frontiers in immunology, 11, 567046-567046 (2021-03-02)
Early childhood infections have been implicated in the development of immune-mediated diseases, such as allergies, asthma, and type 1 diabetes. We set out to investigate the immunomodulatory effects of early viral infections experienced before the age of one year on
Amy D Shapiro et al.
Blood, 131(12), 1301-1310 (2018-01-13)
Congenital plasminogen deficiency is caused by mutations in PLG, the gene coding for production of the zymogen plasminogen, and is an ultrarare disorder associated with abnormal accumulation or growth of fibrin-rich pseudomembranous lesions on mucous membranes. Left untreated, these lesions
Guilherme Guaragna-Filho et al.
Journal of pediatric endocrinology & metabolism : JPEM, 31(2), 191-194 (2018-01-08)
Inhibin B is a hormone produced by the Sertoli cells that can provide important information for the investigation of disorders of sex development (DSD) with 46,XY karyotype. The aim of this study is to compare two enzyme-linked immunosorbent assay (ELISA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service