RAB0508
Human VEGF ELISA Kit
for cell and tissue lysates
Synonym(s):
VEGF-A, Vascular endothelial growth factor A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell lysate
sample type tissue lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 8.23-6000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... VEGFA(7422)
General description
The Human VEGF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human VEGF in cell lysate and tissue lysate.
Immunogen
Recombinant Human VEGF-A
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
VEGF (vascular endothelial growth factor) is a potent angiogenic cytokine. It stimulates proliferation and migration of endothelial cells. VEGF induces microvascular permeability by enhancing function of vesicular-vacuolar organelle (VVO). VEGF plays a prominent role in both normal and pathological angiogenesis. Substantial evidence suggests that VEGF is involved in the induction of tumor metastasis and intraocular neovascular syndromes. Increased expression of VEGF causes retinal angiogenesis, thus, inhibition of VEGF using soluble VEGF-receptor chimeric proteins can be considered as a potential therapy for ischemic retinal disease. Polymorphism in VEGF at C (-634) G is linked with diabetic retinopathy in type 2 diabetes. Altered expression of VEGF is also associated with rheumatoid arthritis, Graves′ disease, psoriasis, cardiovascular disease, and peripheral limb disease.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Miyako Kurihara-Shimomura et al.
International journal of oncology, 56(5), 1093-1100 (2020-04-23)
Mast cells (MCs) are present in the tumor stroma, and MCs that express the mast cell‑specific proteases tryptase and chymase (MCTC) exhibit several tumor‑related functions. It was previously reported that melanoma inhibitory activity (MIA) gene family members, including MIA, MIA2
Piyush Gondaliya et al.
PloS one, 13(11), e0208044-e0208044 (2018-11-30)
The role of DNA methylation has not been enough explored in pathophysiology of diabetic nephropathy (DN). However, according to recent reports it has been inferred that hypermethylation could be one of the principle cause associated with the enhancement of DN.
A common polymorphism in the 5'-untranslated region of the VEGF gene is associated with diabetic retinopathy in type 2 diabetes.
Awata T
Diabetes, 51, 1635-1639 (2002)
Vascular permeability factor/vascular endothelial growth factor: a multifunctional angiogenic cytokine.
Brown LF
EXS, 79, 233-269 (1997)
Suppression of retinal neovascularization in vivo by inhibition of vascular endothelial growth factor (VEGF) using soluble VEGF-receptor chimeric proteins.
Aiello LP
Proceedings of the National Academy of Sciences of the USA, 92, 10457-10461 (1995)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service