Skip to Content
Merck
All Photos(1)

Key Documents

PZ0115

Sigma-Aldrich

CP-31398 dihydrochloride hydrate

≥98% (HPLC)

Synonym(s):

N′-[2-[2-(4-Methoxyphenyl)ethenyl]-4-quinazolinyl]-N,N-dimethyl-1,3-propanediamine dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N4O·2HCl · xH2O
Molecular Weight:
435.39 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

storage condition

desiccated

color

yellow

solubility

H2O: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

O.Cl.Cl.COc1ccc(cc1)\C=C\c2nc(NCCCN(C)C)c3ccccc3n2

InChI

1S/C22H26N4O.2ClH.H2O/c1-26(2)16-6-15-23-22-19-7-4-5-8-20(19)24-21(25-22)14-11-17-9-12-18(27-3)13-10-17;;;/h4-5,7-14H,6,15-16H2,1-3H3,(H,23,24,25);2*1H;1H2/b14-11+;;;

InChI key

JXIVIAMOMIONKY-UWCBQFGESA-N

Gene Information

human ... TP53(7157)
mouse ... TP53(22059)
rat ... TP53(24842)

Related Categories

Application

CP-31398 dihydrochloride hydrate has been used as a p53 stabilizer:
  • to evaluate its effects on the upregulation of miRNA in human neuroblastoma cells
  • to study its effects on arsenic trioxide (ATO) stabilization of p53 folding
  • to study its effects on regulation of miR-34 in PC12 cells

Biochem/physiol Actions

CP-31398 dihyrochloride hydrate is a p53 stabilizer; apoptosis inducer.
CP-31398 is a styryl quinazoline that functions in preserving the activity of p53 as a tumor suppressor and transcription factor. The DNA-binding activity and apoptosis functionality of the p53 are restored by CP-31398. CP-31398 exhibits therapeutic effects against liver, skin, pancreatic, and colon cancers.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William D Johnson et al.
Toxicology, 289(2-3), 141-150 (2011-08-26)
CP-31398 (N'-[2-[2-(4-methoxyphenyl)ethenyl]-4-quinazolinyl]-N,N-dimethyl-1,3-propanediamine dihydrochloride) is a styrylquinazoline that stabilizes the DNA binding conformation of p53, thereby maintaining the activity of p53 as a transcription factor and tumor suppressor. In consideration of the potential use of p53 stabilizers for cancer prevention and
Abhishek Jauhari et al.
Molecular neurobiology, 55(2), 936-945 (2017-01-14)
Differentiation of neural stem cells (NSC's) to mature and functional neurons requires coordinated expression of mRNA, microRNAs (miRNAs) and regulatory proteins. Our earlier unbiased miRNA profiling studies have identified miR-200, miR-34 and miR-221/222 as maximally up-regulated miRNA families in differentiating
Ling Liu et al.
International journal of oncology, 54(3), 942-954 (2019-01-11)
Endometrial cancer (EC) is one of the most common malignancies of the female reproductive system, and metastasis is a major cause of mortality. In this study, we aimed to explore the role of CP‑31398 in the migration, invasion and apoptosis
Abhishek Jauhari et al.
Molecular neurobiology, 54(7), 4986-4995 (2016-08-16)
MicroRNAs (miRNAs) are generated by endonuclease activity of Dicer, which also helps in loading of miRNAs to their target sequences. SH-SY5Y, a human neuroblastoma and a cellular model of neurodevelopment, consistently expresses genes related to neurodegenerative disorders at different biological
Shuo Chen et al.
Cancer cell, 39(2), 225-239 (2020-12-29)
TP53 is the most frequently mutated gene in cancer, yet these mutations remain therapeutically non-actionable. Major challenges in drugging p53 mutations include heterogeneous mechanisms of inactivation and the absence of broadly applicable allosteric sites. Here we report the identification of

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service