Skip to Content
Merck
All Photos(1)

Key Documents

P4743

Sigma-Aldrich

Anti-PUMA/bbc3, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Bcl-2 Binding Component 3, Anti-p53 Upregulated Modulator of Apoptosis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~23 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

microarray: suitable
western blot: 2-4 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BBC3(27113)

Related Categories

General description

p53 upregulated modulator of apoptosis (PUMA) protein is a member of the B-cell lymphoma 2 (BCL2) family. It is a pro-apoptotic BCL-2 homology 3 (BH3)-only protein and has a small BH3 domain. The PUMA gene is mapped on the human chromosome at 19q13.32.

Immunogen

synthetic peptide corresponding to amino acids 2 to 16 of human PUMA-α.

Application

Anti-PUMA/bbc3, N-Terminal antibody produced in rabbit has been used in western blotting and immunoprecipitation.

Biochem/physiol Actions

p53 upregulated modulator of apoptosis (PUMA) acts as a determinant of the levels of germ cells during ovarian development. It promotes apoptosis in various cellular stress insults. Deletion of the PUMA gene is observed in several kinds of human cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Li et al.
Cancer gene therapy, 23(9), 295-302 (2016-08-06)
Oral squamous cell carcinoma (OSCC) is the most common cancer of the head and neck and is associated with a high rate of lymph node metastasis. The initial step in the metastasis and transition of tumors is epithelial-mesenchymal transition (EMT)-induced
Min Li et al.
Pharmaceuticals (Basel, Switzerland), 16(1) (2023-01-22)
Colorectal cancer is one of the most common malignancies, and the topoisomerase inhibitor irinotecan (CPT-11)-based chemotherapeutic regimen is currently the first-line treatment with impressive therapeutic efficacy. However, irinotecan has several clinically significant side effects, including diarrhea, which limit its clinical
Michelle Myers et al.
Reproduction (Cambridge, England), 148(2), 211-219 (2014-05-27)
The number of primordial follicles initially established within the ovary is influenced by the extent of germ cell death during foetal ovarian development, but the mechanisms that mediate this death have not been fully uncovered. In this study, we identified
Bibiana I Ferreira et al.
Haematologica, 93(5), 670-679 (2008-03-28)
Low-grade B-cell lymphomas are a very heterogeneous group of tumors, whose differential diagnosis is frequently compromised by the lack of specific cytogenetic or molecular features. Our objective was to search for genomic features that allow a better molecular identification of
P F Cartron et al.
Cell death & disease, 3, e421-e421 (2012-11-16)
Apoptosis has a crucial role in anti-cancer treatment. The proteins of the BCL-2 family are core members of the apoptotic program. Thus, we postulated that alterations in the expression of BCL-2 protein family, and in particular in that of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service