Skip to Content
Merck
All Photos(2)

Key Documents

P2871

Sigma-Aldrich

Anti-Cytokeratin pan antibody, Mouse monoclonal

clone C-11, purified from hybridoma cell culture

Synonym(s):

Anti-CK-4, Anti-CK4, Anti-CYK4, Anti-K4, Anti-WSN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

C-11, monoclonal

form

buffered aqueous solution

technique(s)

immunocytochemistry: 15-30 μg/mL using methanol/acetone-fixed PTK-2 cells or A549 - human lung carcinoma cells
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 15-30 μg/mL using human skin
immunohistochemistry (frozen sections): suitable
microarray: suitable
western blot: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Monoclonal Anti-Cytokeratin, pan (mouse IgG1 isotype) is derived from the C-11 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a keratin-enriched preparation from cultured human epidermoid carcinoma cell line A431. Cytokeratin is a member of the intermediate filaments family.

Application

Monoclonal Anti-Cytokeratin pan antibody has been used in immunoblotting, dot blot, immunohistochemistry, immunocytochemistry, immunofluorescence, immunostaining, detecting micro-metastases in lymph nodes, bone marrow, and other tissues and for determining the origin of poorly differentiated tumors.

Biochem/physiol Actions

Cytokeratin plays a major role in observing carcinomas.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatio-temporal expression of MMP-2, MMP-9 and tissue kallikrein in uteroplacental units of the pregnant guinea-pig (Cavia porcellus)
Corthorn J, et al.
Reproductive Biology and Endocrinology, 5(1), 27-27 (2007)
Caspase-cleaved cytokeratin 18 fragment (M30) as marker of postoperative residual tumor load in colon cancer patients
Ausch C, et al.
European Journal of Surgical Oncology, 35(11), 1164-1168 (2009)
Gloria Valdés et al.
Reproductive sciences (Thousand Oaks, Calif.), 27(8), 1648-1655 (2020-05-21)
The pleiotropic kininogen-kallikrein-kinin system is upregulated in pregnancy and localizes in the uteroplacental unit. To identify the systemic and local participation of the bradykinin type 2 receptor (B2R), this was antagonized by Bradyzide (BDZ) during 2 periods: from days 20
Isabel Egaña et al.
Scientific reports, 7(1), 15453-15453 (2017-11-15)
Paladin (Pald1, mKIAA1274 or x99384) was identified in screens for vascular-specific genes and is a putative phosphatase. Paladin has also been proposed to be involved in various biological processes such as insulin signaling, innate immunity and neural crest migration. To
Co-culture with endometrial stromal cells enhances the differentiation of human embryonic stem cells into endometrium-like cells
Yu W, et al.
Experimental and Therapeutic Medicine, 10(1), 43-50 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service