Skip to Content
Merck
All Photos(1)

Key Documents

O7885

Sigma-Aldrich

Okadaic acid potassium salt from Prorocentrum concavum

≥90% (HPLC), powder

Synonym(s):

OA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H67KO13
CAS Number:
Molecular Weight:
843.09
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

plant (Prorocentrum concavum)

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white

solubility

H2O: 0.1 mg/mL
DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

[K+].CC(CC(O)C1OC2CCC3(CCC(C)(O3)\C=C\C(C)C4CC(C)=CC5(O4)OC(C)(CCC5O)CC(C)(O)C([O-])=O)OC2C(O)C1=C)C6CC7(CCCOC7)COC6C

InChI

1S/C46H72O13.K/c1-27-20-35(56-46(22-27)36(48)12-15-42(7,59-46)24-43(8,52)40(50)51)28(2)10-14-41(6)17-18-45(58-41)16-11-34-39(57-45)37(49)30(4)38(55-34)33(47)21-29(3)32-23-44(26-54-31(32)5)13-9-19-53-25-44;/h10,14,22,28-29,31-39,47-49,52H,4,9,11-13,15-21,23-26H2,1-3,5-8H3,(H,50,51);/q;+1/p-1/b14-10+;

InChI key

CWKZDWKRECTBCU-KMZJGFRYSA-M

Biochem/physiol Actions

Dinoflagellate toxin and an ionophore-like polyether derivative of a 38 carbon, fatty acid. Readily enters cells. Inhibitor of type 1 and type 2A protein phosphatases. Does not inhibit tyrosine phosphatases, alkaline phosphatases or acid phosphatase. Known tumor promotor. Used to study various cellular processes including cell cycle, apoptosis, nitric oxide metabolism and calcium signaling.
Okadaic acid is a dinoflagellate toxin and an ionophore-like polyether derivative of a 38 carbon, fatty acid.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulan Zhao et al.
Life sciences, 246, 117382-117382 (2020-02-01)
Our preliminary research revealed that metformin, a classic anti-diabetic drug, could rescue Parkin protein expression and mitophagy in high glucose-challenged human renal epithelial cells in vitro, but the molecular mechanism remains to be explored. In the study, Human Renal Cortical
Mitchell J George et al.
International journal of molecular sciences, 22(12) (2021-07-03)
Myosin Light Chain (MLC) regulates platelet contraction through its phosphorylation by Myosin Light Chain Kinase (MLCK) or dephosphorylation by Myosin Light Chain Phosphatase (MLCP). The correlation between platelet contraction force and levels of MLC phosphorylation is unknown. We investigate the
Yongqiang Chen et al.
Frontiers in cell and developmental biology, 10, 823251-823251 (2022-03-22)
Glioblastoma (GBM) is the most common and aggressive type of brain cancer in adults, with temozolomide (TMZ) being widely used as the standard chemotherapy drug for its treatment. However, GBM frequently becomes resistant to TMZ treatment due to various mechanisms
Huijing Yin et al.
Nature communications, 12(1), 4230-4230 (2021-07-11)
Extracellular matrix protein-1 (ECM1) promotes tumorigenesis in multiple organs but the mechanisms associated to ECM1 isoform subtypes have yet to be clarified. We report in this study that the secretory ECM1a isoform induces tumorigenesis through the GPR motif binding to
Yongqiang Chen et al.
Autophagy, 18(6), 1274-1296 (2021-09-18)
Cancer cell growth is dependent upon the sustainability of proliferative signaling and resisting cell death. Macroautophagy/autophagy promotes cancer cell growth by providing nutrients to cells and preventing cell death. This is in contrast to autophagy promoting cell death under some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service