Skip to Content
Merck
All Photos(2)

Key Documents

N7782

Sigma-Aldrich

Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-iNOS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 130 kDa

species reactivity

rat, mouse

technique(s)

microarray: suitable
western blot: 1:10,000 using cells extract of murine RAW 264.7 macrophages activated with LPS and IFN-γ.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Nos2(18126)
rat ... Nos2(24599)

General description

Anti-Nitric Oxide Synthase, inducible is specific for nitric oxide synthase (NOS), derived from activated mouse macrophage RAW 264.7 cells (iNOS, 130 kDa), but does not recognize NOS derived from brain tissue (bNOS) and endothelial cells (eNOS). The NOS isoform characterized in macrophages is a 130 kDa protein, also known as macrophage NOS (macNOS; mNOS), NOS2, or inducible NOS (iNOS). iNOS is an interferon (IFN)-γ-inducible protein. This gene is located on human chromosome 17q11.

Specificity

Anti-Nitric Oxide Synthase, Inducible is specific for nitric oxide synthase (NOS), derived from activated mouse macrophage RAW 264.7 cells (iNOS, 130 kDa), but does not recognize NOS derived from brain tissue (bNOS) and endothelial cells (eNOS). By immunoblotting of an activated mouse macrophage RAW 264.7 cells extract, the staining of the iNOS band is specifically inhibited by the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 1126-1144 of nitric oxide synthase (NOS) of mouse macrophage origin (iNOS, also termed macNOS) conjugated to KLH. The immunogen sequence is highly conserved in rat iNOS and diverges from the human iNOS.

Application

Anti-Nitric Oxide Synthase, Inducible antibody produced in rabbit has been used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Inducible nitric oxide synthase (NOS2) produces nitric oxide (NO), which is expected to be lethal to malaria parasites in vitro.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Bach et al.
Journal of neurochemistry, 78(1), 109-120 (2001-07-04)
One of the pathophysiological features of Alzheimer's disease is astrocytosis around senile plaques. Reactive astrocytes may produce proinflammatory mediators, nitric oxide, and subsequent reactive oxygen intermediates such as peroxynitrites. In the present study, we investigated the possible role of the
Savita Verma et al.
Frontiers in immunology, 8, 658-658 (2017-06-27)
Pneumonitis and pulmonary fibrosis are predominant consequences of radiation exposure, whether planned or accidental. The present study, demonstrates radioprotective potential of a formulation, prepared by combining podophyllotoxin and rutin (G-003M), in mice exposed to 11 Gy thoracic gamma radiation (TGR). Treated
Sung-Suk Suh et al.
International journal of medical sciences, 15(9), 929-936 (2018-07-17)
Inflammation mediated by the innate immune system is an organism's protective mechanism against infectious environmental risk factors. It is also a driver of the pathogeneses of various human diseases, including cancer development and progression. Microalgae are increasingly being focused on
Heterotrimeric G protein-dependent WNT-5A signaling to ERK1/2 mediates distinct aspects of microglia proinflammatory transformation
Halleskog C, et al.
Journal of Neuroinflammation, 9(1), 111-111 (2012)
Inducible nitric oxide synthase 2 promoter polymorphism and malaria disease severity in children in Southern Ghana
Dzodzomenyo M, et al.
PLoS ONE, 13(8), e0202218-e0202218 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service