Skip to Content
Merck
All Photos(2)

Key Documents

N3889

Sigma-Aldrich

Nitrazepam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O3
CAS Number:
Molecular Weight:
281.27
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

SMILES string

[O-][N+](=O)c1ccc2NC(=O)CN=C(c3ccccc3)c2c1

InChI

1S/C15H11N3O3/c19-14-9-16-15(10-4-2-1-3-5-10)12-8-11(18(20)21)6-7-13(12)17-14/h1-8H,9H2,(H,17,19)

InChI key

KJONHKAYOJNZEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hypnotic; ligand for the GABAA receptor benzodiazepine modulatory site.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N Inazu et al.
Biochemical pharmacology, 52(10), 1585-1591 (1996-11-22)
We investigated the effects of minor and major tranquilizers on ovarian and adrenal aldo-keto reductase activity towards five substrates in relation to ovulation in mature cycling rats. Nitrazepam (NZP) did not alter ovarian and adrenal weights or body weight, although
J Liu et al.
Journal of pharmaceutical sciences, 87(12), 1632-1638 (1999-04-03)
The purpose of this research was to investigate the influence of surfactants on the dissolution of benzodiazepines in interactive mixtures. The dissolution of ternary interactive mixtures consisting of micronized drugs (oxazepam, nitrazepam, and flunitrazepam) and micronized surfactants (sodium lauryl sulfate
Mihalj Poša et al.
Colloids and surfaces. B, Biointerfaces, 86(2), 285-291 (2011-05-07)
The formation of mixed micelles built of 7,12-dioxolithocholic and the following hydrophobic bile acids was examined by conductometric method: cholic (C), deoxycholic (D), chenodeoxycholic (CD), 12-oxolithocholic (12-oxoL), 7-oxolithocholic (7-oxoL), ursodeoxycholic (UD) and hiodeoxycholic (HD). Interaction parameter (β) in the studied
Masatomo Suetsugi et al.
Progress in neuro-psychopharmacology & biological psychiatry, 31(4), 839-847 (2007-03-03)
The first-night effect is a well-known phenomenon that is considered to result from a subject's lack of adaptation to the unfamiliar environment of a sleep laboratory and to the technical equipment used for polysomnography. The effect has been explored as
Mihalj Posa et al.
Colloids and surfaces. B, Biointerfaces, 74(1), 84-90 (2009-07-28)
Critical micellear concentrations (CMC) were determined for two novel promoters of membrane permeability-7-monoketocholic acid (7-MKC) and 12-monoketocholic acid (12-MKC), using two non-invasive ((1)H NMR relaxation experiment and conductometry) and two invasive (spectral shift and partition coefficient of the probe molecule)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service