Skip to Content
Merck
All Photos(1)

Key Documents

N170

Sigma-Aldrich

NS 1619

powder

Synonym(s):

1,3-Dihydro-1-[2-hydroxy-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-2H-benzimidazol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8F6N2O2
CAS Number:
Molecular Weight:
362.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

powder

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.47 mg/mL
ethanol: 26.6 mg/mL
0.1 M NaOH: 5.2 mg/mL
DMSO: 5.3 mg/mL
H2O: insoluble

SMILES string

Oc1ccc(cc1N2C(=O)Nc3cc(ccc23)C(F)(F)F)C(F)(F)F

InChI

1S/C15H8F6N2O2/c16-14(17,18)7-1-3-10-9(5-7)22-13(25)23(10)11-6-8(15(19,20)21)2-4-12(11)24/h1-6,24H,(H,22,25)

InChI key

YLFMCMWKHSDUCT-UHFFFAOYSA-N

Gene Information

Application

NS 1619 has been used as calcium-activated potassium channels (BK)-activator: to study its vaso-relaxing effects on the thoracic aortic rings from mice, to evaluate its effects on the sarco-/endoplasmic reticulum Ca2+-ATPase (SERCA) activity from skeletal muscles, to determine its effects on the expression of caveolin-1 protein in mice brain tumor microvascular endothelial cells

Biochem/physiol Actions

NS 1619 is a selective large conductance Ca2+-activated K+-channel activator. The compound is known for its vasodilating and anti-proliferative properties. It is studied that NS 1619 can protect cardiac muscle against ischemia and reperfusion injury.
NS-1619 can promote the formation of mitochondrial reactive oxygen species (ROS) in cardiac myocytes. It enhances the formation of ROS in cultured brain slices. NS-1619 has the ability to enhance HO-1 activity in wild-type (WT) animals.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincenzo Calderone et al.
Naunyn-Schmiedeberg's archives of pharmacology, 375(1), 73-80 (2007-01-05)
Large-conductance calcium-activated potassium channels (BK) of smooth muscle play a role in the relevant modulation of vascular tone, due to their calcium- and voltage-dependent mechanisms of activation. A potential role of endothelial BK channels has also been suggested by approaches
Yajun Liu et al.
American journal of physiology. Gastrointestinal and liver physiology, 302(1), G44-G54 (2011-09-17)
The objectives of this study were to determine whether neutrophil depletion with anti-neutrophil serum (ANS) or preconditioning with the hydrogen sulfide (H(2)S) donor NaHS (NaHS-PC) 24 h prior to ischemia-reperfusion (I/R) would prevent postischemic mitochondrial dysfunction in rat intestinal mucosa
Rupal P Soder et al.
European journal of pharmacology, 670(1), 252-259 (2011-09-15)
Large conductance voltage- and Ca(2+)-activated K(+) (BK) channels are important in regulating detrusor smooth muscle (DSM) function. Here, we examined systematically how the BK channel pharmacological activation modulates DSM contractility. NS1619, a potent BK channel activator, was utilized as a
Antoni Wrzosek
Cell calcium, 56(1), 14-24 (2014-05-13)
NS1619 (1,3-dihydro-1-[2-hydroxy-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-2H-benzimidazole-2-one) is widely used as a large-conductance Ca(2+)-activated K(+) (BKCa) channel opener. It was previously reported that activation of BKCa channels by NS1619 could protect the cardiac muscle against ischaemia and reperfusion injury. This study reports the effects of
Rui-Ping Cai et al.
Journal of the neurological sciences, 369, 109-118 (2016-09-23)
NS1619, a calcium-activated potassium channel (Kca channel) activator, can selectively and time-dependently accelerate the formation of transport vesicles in both the brain tumor capillary endothelium and tumor cells within 15min of treatment and then increase the permeability of the blood-brain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service