L2025
Luria Agar Base, Miller
Tryptone, yeast extract, sodium chloride and agar
Synonym(s):
LA, modified, Luria-Bertani media, modified, Lysogeny broth, modified
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41106200
eCl@ss:
42040102
NACRES:
NA.72
Recommended Products
Quality Level
form
powder
technique(s)
microbiological culture: suitable
application(s)
agriculture
microbiology
General description
Luria Bertani (LB) medium is one of the complex media that is widely used to grow Escherichia coli in small-scale cultures.
Application
Luria Agar Base, Miller has been used as a growth medium during plasmid amplification. It has also been used as a culture medium for strains of Penicillium sp. laika.
Luria broth/agar (LA), Miller (L1900) is a bacteria growth mixture of sodium chloride (0.5g/L); tryptone (10g/L) yeast extract (5g/L) and agar (15g/L) used to culture members of the Enterobacteriaceae family and for coliphage plaque assays.
Components
Contains: Tryptone, yeast extract, sodium chloride and agar.
Reconstitution
Recommended use at 30.5 g per liter.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Daria Julkowska et al.
Journal of bacteriology, 187(1), 65-76 (2004-12-17)
The natural wild-type Bacillus subtilis strain 3610 swarms rapidly on the synthetic B medium in symmetrical concentric waves of branched dendritic patterns. In a comparison of the behavior of the laboratory strain 168 (trp) on different media with that of
C Niu et al.
Applied and environmental microbiology, 70(12), 6951-6956 (2004-12-03)
The specific biofilm formation (SBF) assay, a technique based on crystal violet staining, was developed to locate plant essential oils and their components that affect biofilm formation. SBF analysis determined that cinnamon, cassia, and citronella oils differentially affected growth-normalized biofilm
Mario Losen et al.
Biotechnology progress, 20(4), 1062-1068 (2004-08-07)
Shake-flask cultures are widely used for screening of high producing strains. To select suitable strains for production scale, cultivation parameters should be applied that provide optimal growth conditions. A novel method of measuring respiratory activity in shake-flask cultures was employed
Anthony S Danko et al.
Applied and environmental microbiology, 70(10), 6092-6097 (2004-10-07)
Pseudomonas putida strain AJ and Ochrobactrum strain TD were isolated from hazardous waste sites based on their ability to use vinyl chloride (VC) as the sole source of carbon and energy under aerobic conditions. Strains AJ and TD also use
Chujin Ruan et al.
Current biology : CB, 32(24), 5285-5294 (2022-12-02)
The amount of bacterial diversity present on many surfaces is enormous; however, how these levels of diversity persist in the face of the purifying processes that occur as bacterial communities expand across space (referred to here as range expansion) remains
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service