Skip to Content
Merck
All Photos(1)

Documents

K3769

Sigma-Aldrich

KW-3902

≥98% (HPLC)

Synonym(s):

1,3-dipropyl-8-(3-noradamantyl)xanthine, 8-(Hexahydro-2,5-methanopentalen-3a(1H)-yl)-3,7-dihydro-1,3-dipropyl-1H-purine-2,6-dione, MK-7418, Rolofylline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N4O2
CAS Number:
Molecular Weight:
356.46
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 20 mg/mL
soluble

storage temp.

2-8°C

SMILES string

CCCN1C(=O)N(CCC)c2[nH]c(nc2C1=O)[C@]34C[C@H]5C[C@H](C[C@@H]3C5)C4

InChI

1S/C20H28N4O2/c1-3-5-23-16-15(17(25)24(6-4-2)19(23)26)21-18(22-16)20-10-12-7-13(11-20)9-14(20)8-12/h12-14H,3-11H2,1-2H3,(H,21,22)/t12-,13+,14-,20-

InChI key

PJBFVWGQFLYWCB-OYEQCZOJSA-N

Biochem/physiol Actions

KW-3902 is an A1 adenosine receptor antagonist and is over 800 fold more selective for an A1 receptor versus the A2A receptor.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mustafa M Dohadwala et al.
Cardiovascular therapeutics, 26(4), 276-286 (2008-11-28)
Acute decompensated heart failure (ADHF), generally related to signs and symptoms of volume overload, is one the most common reasons for hospitalization in the United States. Recently, it has been observed that the majority of patients with ADHF have baseline
Piotr Ponikowski et al.
European journal of heart failure, 12(11), 1238-1246 (2010-09-09)
The direct effects of adenosine A1 receptor antagonists on haemodynamic parameters in patients with acute heart failure (HF) remain largely unknown. We evaluated the haemodynamic effects of the AA(1)RA rolofylline in 59 HF patients with concomitant renal impairment (estimated creatinine
Adriaan A Voors et al.
Journal of the American College of Cardiology, 57(19), 1899-1907 (2011-05-07)
This study sought to assess the effects of rolofylline on renal function in patients with acute heart failure (AHF) and renal dysfunction randomized in PROTECT (Placebo-Controlled Randomized Study of the Selective A(1) Adenosine Receptor Antagonist Rolofylline for Patients Hospitalized With
Marco Metra et al.
European journal of heart failure, 12(5), 499-507 (2010-03-17)
Dyspnoea is the most common symptom leading to hospitalization for acute heart failure (AHF). Its early and persistent relief is an important goal of therapy, but little is known about its course, determinants, and prognostic significance. In a post hoc
Gad Cotter et al.
Journal of cardiac failure, 14(8), 631-640 (2008-10-18)
Rolofylline, an adenosine A(1) receptor antagonist, facilitates diuresis and preserves renal function in patients with acute heart failure (AHF) with renal impairment. Although not powered around any specific hypothesis, this pilot study was designed to identify an efficacious dose while

Articles

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service