Skip to Content
Merck
All Photos(1)

Key Documents

I5273

Sigma-Aldrich

Interleukin-5 human

recombinant, expressed in baculovirus infected Sf21 cells, ≥97% (SDS-PAGE), lyophilized powder, suitable for cell culture

Synonym(s):

IL-5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.1-0.5 ng/mL EC50

mol wt

32-34 kDa

packaging

pkg of 5 μg

concentration

0.5-10 ng/mL

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL5(3567)

Biochem/physiol Actions

IL-5 (interleukin-5) is a maturation and differentiation factor for eosinophils. Thus, it is associated with allergy and related disorders. IL-5 association with its receptor causes activation of MEK/ERK (mitogen activated protein kinase kinase/extracellular-regulated kinase), p38 MAPK (mitogen activated protein kinase) and JAK/STAT (Janus kinase/signal transducer and activator of transcription) pathways.
Interleukin-5 is a product of activated T lymphocytes and exhibits activity on eosinophils, B cells and thymocytes. Synonyms or biological activities attributed to IL-5 include: EDF (eosinophil differentiating factor), BCGF-II (B cell growth factor-II), TRF (T cell replacing factor), IgA-EF (IgA enhancing factor) and EO-CSF (eosinophil colony stimulating factor). Natural IL-5 is a 45-50 kDa dimeric glycoprotein, although glycosylation is not required for activity. Human and mouse IL-5 exhibit homology at the nucleotide and amino acid levels and show species cross-reactivity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.1 mg bovine serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Silveira et al.
Atherosclerosis, 239(1), 125-130 (2015-01-15)
Genetic variants robustly associated with coronary artery disease were reported in the vicinity of the interleukin (IL)-5 locus, and animal studies suggested a protective role for IL-5 in atherosclerosis. Therefore, we set this work to explore IL-5 as a plasma
Takehito Kobayashi et al.
Allergology international : official journal of the Japanese Society of Allergology, 64 Suppl, S30-S36 (2015-09-08)
Eosinophils recognize various stimuli, such as cytokines, chemokines, immunoglobulins, complement, and external pathogens, resulting in their accumulation in mucosal tissues and the progression of inflammation. Eosinophils are also involved in innate Th2-type immune responses mediated through endogenous danger signals, including
Mandy E Burnham et al.
PloS one, 9(8), e103122-e103122 (2014-08-15)
Eosinophils function contributes to human allergic and autoimmune diseases, many of which currently lack curative treatment. Development of more effective treatments for eosinophil-related diseases requires expanded understanding of eosinophil signaling and biology. Cell signaling requires integration of extracellular signals with
Ma Paz Zafra et al.
International journal of molecular sciences, 16(3), 5434-5451 (2015-03-13)
Eosinophils are one of the key inflammatory cells in asthma. Eosinophils can exert a wide variety of actions through expression and secretion of multiple molecules. Previously, we have demonstrated that eosinophils purified from peripheral blood from asthma patients express high

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service