Skip to Content
Merck
All Photos(1)

Documents

I3650

Sigma-Aldrich

Interleukin-4 from rat

recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)

Synonym(s):

rIL-4, IL-4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

mol wt

predicted mol wt 13.5 kDa

packaging

pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Il4(287287)

Biochem/physiol Actions

Interleukin-4 is a lymphokine with profound effects on the growth and differentiation of immunologically competent cells. IL-4 is also known as B cell stimulatory factor-1 (BSF-1), T cell growth factor-2 (TCGF-2) and mast-cell growth factor-2 (MCGF-2). Inhibits VEGF-induced and bFGF-induced angiogenesis. IL-4 is a complex glycoprotein released by a subset of activated T cells. Human and mouse IL-4 share 50% amino acid sequence homology, but their biological actions are species-specific.
Interleukin-4, a lymphokine with profound effects on the growth and differentiation of immunologically competent cells, inhibits VEGF-induced and bFGF-induced angiogenesis. Human and mouse IL-4 share 50% amino acid sequence identity, but their biological actions are species-specific.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.25 mg bovine serum albumin.

Analysis Note

The biological activity of recombinant rat IL-4 was measured by its ability to stimulate proliferation of rat splenocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Nunan et al.
Glia, 62(8), 1313-1327 (2014-05-08)
Neurogenesis, the production of new neurons from neural stem/progenitor cells (NSPCs), occurs throughout adulthood in the dentate gyrus of the hippocampus, where it supports learning and memory. The innate and adaptive immune systems are increasingly recognized as important modulators of
Monique Michels et al.
Clinical science (London, England : 1979), 134(7), 765-776 (2020-03-29)
In order to modulate microglial phenotypes in vivo, M1 microglia were depleted by administration of gadolinium chloride and the expression of M2 microglia was induced by IL-4 administration in an animal model of sepsis to better characterize the role of
Jeremy A Daniel et al.
The Journal of cell biology, 183(5), 777-783 (2008-12-03)
Cellular responses to both physiological and pathological DNA double-strand breaks are initiated through activation of the evolutionarily conserved ataxia telangiectasia mutated (ATM) kinase. Upon DNA damage, an activation mechanism involving autophosphorylation has been reported to allow ATM to phosphorylate downstream
Mingming Zhang et al.
PloS one, 9(10), e109146-e109146 (2014-10-03)
It has been shown that Faecalibacterium prausnitzii (F. prausnitzii), one of the dominant intestinal bacterial flora, may protect colonic mucosa against the development of inflammation and subsequent inflammatory bowel disease (IBD), with the underlying mechanisms being unclear. The impacts of
Rebecca Winkelmann et al.
PloS one, 9(5), e97953-e97953 (2014-05-31)
Maturation as well as antigen-dependent activation of B cells is accompanied by alternating phases of proliferation and quiescence. We and others have previously shown that Krüppel-like factor 2 (KLF2), a regulator of T cell quiescence and migration, is upregulated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service