Skip to Content
Merck
All Photos(12)

Documents

HPA025770

Sigma-Aldrich

Anti-FH antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Anti-fumarase, Anti-fumarate hydratase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DASVSFTENCVVGIQANTERINKLMNESLMLVTALNPHIGYDKAAKIAKTAHKNGSTLKETAIELGYLTAEQFDEWVKPK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FH(2271)

General description

The gene FH (fumarate hydratase) is mapped to human chromosome 1q42.3-q43. The encoded protein is responsible for the conversion of fumarate to malate in the TCA (tricarboxylic acid) cycle. Fumarate is an oncometabolite and mutation in the FH gene causes accumulation of fumarate. Mutations in this gene are associated with HLRCC (hereditary leiomyomatosis and renal cell cancer) syndrome. Some mutations also cause fumarase deficiency resulting in neurological impairment.

Immunogen

fumarate hydratase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76561

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kentaro Suda et al.
The Journal of clinical endocrinology and metabolism, 105(6) (2020-04-07)
Germline mutations in fumarate hydratase (FH) gene are known to cause hereditary leiomyomatosis and renal cell carcinoma (HLRCC) and are occasionally accompanied with cutaneous and uterine leiomyoma or cortisol-producing adrenocortical hyperplasia. However, the association between FH mutations and cardiac or
Genetic variations in genes of metabolic enzymes predict postoperational prognosis of patients with colorectal cancer.
Dong G
Molecular Cancer, 14, 171-171 (2015)
Molecular analysis and prenatal diagnosis of human fumarase deficiency.
Coughlin EM
Molecular Genetics and Metabolism, 63, 254-262 (1998)
Fumarate Hydratase-deficient Uterine Leiomyomas Occur in Both the Syndromic and Sporadic Settings.
Harrison WJ, et al.
American Journal of Surgical Pathology, 40, 599-607 (2016)
Xujun Liu et al.
Signal transduction and targeted therapy, 6(1), 375-375 (2021-11-04)
The scope and variety of the metabolic intermediates from the mitochondrial tricarboxylic acid (TCA) cycle that are engaged in epigenetic regulation of the chromatin function in the nucleus raise an outstanding question about how timely and precise supply/consumption of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service