Skip to Content
Merck
All Photos(6)

Documents

HPA018322

Sigma-Aldrich

Anti-TOX antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Thymocyte selection-associated high mobility group box protein TOX, Anti-Thymus high mobility group box protein TOX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PDAPCLGPSPCLDPYYCNKFDGENMYMSMTEPSQDYVPASQSYPGPSLESEDFNIPPITPPSLPDHSLVHLNEVESGYHSLCHPMNHNGLLPFHPQN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TOX(9760)

General description

The gene thymocyte selection-associated high mobility group box protein (TOX) is mapped to human chromosome 8q12.1. TOX belongs to HMG (high-mobility group) box family of DNA-binding proteins. It is mainly expressed in the thymus.

Immunogen

Thymocyte selection-associated high mobility group box protein TOX recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Thymocyte selection-associated high mobility group box protein (TOX) is important for T-cell selection, differentiation and maturation. TOX is up-regulated by pre-T cell receptor (TCR) and TCR activation of immature thymocytes. Presence of TOX results in expanded CD8+ and reduced CD4+ single positive thymocyte subpopulation. TOX promoter is hypermethylated in lung and breast cancer lines, resulting in down-regulation of TOX. However, down-regulation of TOX does not affect the proliferation or migration potential of the cells. TOX is required for IL (Interleukin)-15-mediated natural killer (NK) cell differentiation. In addition, TOX affects the expression of T-bet (T-box expressed in T cells) which plays important role in NK differentiation and maturation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73801

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abstracts from USCAP 2021: Hematopathology (664-756).
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 34(Suppl 2), 927-1049 (2021-03-13)
J Calvani et al.
The British journal of dermatology, 185(2), 419-427 (2021-01-06)
The prognosis of Sézary syndrome (SS) and mycosis fungoides (MF) depends on lymph node (LN) involvement. The usefulness of LN image-guided core-needle biopsies (CNBs), instead of surgical sampling, has been poorly evaluated. To determine the prognostic value of LN CNB
Differential epigenetic regulation of TOX subfamily high mobility group box genes in lung and breast cancers.
Tessema M
PLoS ONE, 7, e34850-e34850 (2012)
Lihi Atzmony et al.
Acta dermato-venereologica, 100(15), adv00230-adv00230 (2020-06-20)
Recent studies suggest that folliculotropic mycosis fungoides (FMF), the most common variant of mycosis fungoides (MF), presents with 2 distinct clinicopathological stages: early indolent stage and more aggressive advanced/tumour stage. To further characterize these stages, miR-155 expression was studied with
A M R Schrader et al.
The British journal of dermatology, 175(2), 382-386 (2016-03-05)
TOX (thymocyte selection-associated high-mobility group box) was shown to be aberrantly expressed in mycosis fungoides (MF) and Sézary syndrome (SS) and is suggested to have additional diagnostic value. However, data on expression in other types of cutaneous T-cell lymphoma (CTCL)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service