Skip to Content
Merck
All Photos(6)

Key Documents

HPA009433

Sigma-Aldrich

Anti-CANX antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Calnexin precursor antibody produced in rabbit, Anti-IP90 antibody produced in rabbit, Anti-Major histocompatibility complex class I antigen-binding protein p88 antibody produced in rabbit, Anti-p90 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DVIDIEDDLDDVIEEVEDSKPDTTAPPSSPKVTYKAPVPTGEVYFADSFDRGTLSGWILSKAKKDDTDDEI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CANX(821)

Looking for similar products? Visit Product Comparison Guide

General description

CANX (calnexin) is an endoplasmic reticulum (ER) chaperone, which is a homolog of calreticulin (Crt). It is an ER-membrane protein with a molecular weight of 67kDa. Its N and P domains face the ER lumen and its C-terminal faces the cytoplasmic region. It is a type I transmembrane protein. Cleavage of this protein results in separation of membrane and cytosolic regions, giving rise to a soluble form found in placenta.

Immunogen

Calnexin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CANX (calnexin) is a chaperone protein, which controls protein folding and quality control. It is specific to proteins composed of monoglucosylated oligosaccharides. It is also responsible for the assembly of viral proteins. The expression of this protein reduces with age, for instance oxidative stress-induced expression in human diploid fibroblasts (HDF) in older population. Thus, it might be involved in conferring protection to cells in various age-related disorders. CANX is responsible for glycan-independent interaction and retention of mutated Gas3/PMP22 (growth arrest-specific protein 3/peripheral myelin protein 22) proteins involved in Charcot-Marie-Tooth 1A and Dejerine Sottas syndrome.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71909

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bok-Hee Choi et al.
Experimental & molecular medicine, 36(5), 499-503 (2004-11-24)
Aging is accompanied by the changes in the cells that decrease their capacity to respond to various forms of stress. Cells are known to respond to stresses through expression of stress-response proteins, heat-shock proteins composed of molecular chaperones. Recent studies
Alessandra Fontanini et al.
The Journal of biological chemistry, 280(3), 2378-2387 (2004-11-13)
Missense point mutations in Gas3/PMP22 are responsible for the peripheral neuropathies Charcot-Marie-Tooth 1A and Dejerine Sottas syndrome. These mutations induce protein misfolding with the consequent accumulation of the proteins in the endoplasmic reticulum and the formation of aggresomes. During folding
Vilasack Thammavongsa et al.
The Journal of biological chemistry, 280(39), 33497-33505 (2005-08-03)
Calnexin is an endoplasmic reticulum chaperone that binds to substrates containing monoglucosylated oligosaccharides. Whether calnexin can also directly recognize polypeptide components of substrates is controversial. We found that calnexin displayed significant conformational lability for a chaperone and that heat treatment
Johan Nordholm et al.
The Journal of cell biology, 216(8), 2283-2293 (2017-07-12)
A primary function of 5' regions in many secretory protein mRNAs is to encode an endoplasmic reticulum (ER) targeting sequence. In this study, we show how the regions coding for the ER-targeting sequences of the influenza glycoproteins NA and HA
Dorthe T Olsen et al.
Protein expression and purification, 92(1), 105-111 (2013-09-24)
Calreticulin (Crt) and calnexin (Cnx) are homologous endoplasmic reticulum (ER) chaperones involved in protein folding and quality control. Crt is a soluble ER luminal Mr 46 kDa protein and Cnx is a Mr 67kDa ER membrane protein. During purification of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service