Skip to Content
Merck
All Photos(6)

Key Documents

HPA008999

Sigma-Aldrich

Anti-EFNB2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EPH-related receptor tyrosine kinase ligand 5 antibody produced in rabbit, Anti-Ephrin-B2 precursor antibody produced in rabbit, Anti-HTK ligand antibody produced in rabbit, Anti-HTK-L antibody produced in rabbit, Anti-LERK-5 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YRRRHRKHSPQHTTTLSLSTLATPKRSGNNNGSEPSDIIIPLRTADSVFCPHYEKVSGDYGHPVYIVQEMP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFNB2(1948)

General description

EFNB2 (ephrin-B2) is a member of the B-class ephrins, which function as ligands for EphB RTKs (receptor tyrosine kinase). It is a transmembrane protein. It is a cell surface signaling glycoprotein, which has a ubiquitous expression pattern. EFNB2 is expressed in mesenchymal cells, with the highest expression in fibroblast. EFNB2 gene is located on human chromosome 13q33.3.

Immunogen

Ephrin-B2 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-EFNB2 antibody produced in rabbit has been used in:
  • immunohistochemistry (1:100)
  • western blot analysis (1:1000)
  • immunostaining

Anti-EFNB2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Ephrin-B2 (EFNB2) regulates sprouting and proliferation of endothelial cells in the endothelium of the developing vasculature. This is in turn, linked with vascular endothelial growth factor (VEGF) receptor endocytosis and signaling. It is crucial in supporting pericytes and vascular smooth muscle cells (VSMCs). It also functions as the regulator of the distribution, internalization and signaling of platelet-derived growth factor receptor β (PDGFRβ) in VSMCs. In castration resistant prostate cancer (CRPC), the expression of this gene is directly suppressed by miR-582-5p. Therefore, it might function as a tumor suppressor functioning through EphB4, in cancer cells expressing EphB4. This protein is responsible for the induction of neuronal differentiation of neural stem cells (NSCs), when presented by neighbouring astrocytes. EFNB2 participates in the development of the cardiovascular system, postnatal angiogenesis, and cardiac remodeling. It serves as a modulator in transforming growth factor-ß (TGF-ß)-mediated organ fibrosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71886

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Essential roles of EphrinB2 in mammalian heart: from development to diseases
Su S-an, et al.
Cell communication and signaling : CCS (2019)
Atsushi Maeno et al.
The Prostate, 74(16), 1604-1612 (2014-09-02)
MicroRNAs are noncoding small RNA that negatively regulate target gene expression by binding to the 3'-UTR of mRNA. Previous studies have shown that several microRNAs play a pivotal role in prostate cancer by acting as oncogenes or tumor suppressors. This
Benhur Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(17), E2156-E2165 (2015-04-01)
The discovery of African henipaviruses (HNVs) related to pathogenic Hendra virus (HeV) and Nipah virus (NiV) from Southeast Asia and Australia presents an open-ended health risk. Cell receptor use by emerging African HNVs at the stage of host-cell entry is
Katie Pricola Fehnel et al.
Experimental & molecular medicine, 52(4), 658-671 (2020-04-15)
We investigated (1) EphrinB2 and EphB4 receptor expression in cerebral AVMs, (2) the impact of an altered EphrinB2:EphB4 ratio on brain endothelial cell function and (3) potential translational applications of these data. The following parameters were compared between AVM endothelial
Cvetan Popov et al.
Frontiers in aging neuroscience, 7, 246-246 (2016-01-19)
Tendon tissues, due to their composition and function, are prone to suffer age-related degeneration and diseases as well as to respond poorly to current repair strategies. It has been suggested that local stem cells, named tendon stem/progenitor cells (TSPCs), play

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service