Skip to Content
Merck
All Photos(6)

Documents

HPA006564

Sigma-Aldrich

Anti-PRKCA antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PKCA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RTTRNDFMGSLSFGVSELMKMPASGWYKLLNQEEGEYYNVPIPEGDEEGNMELRQKFEKAKLGPAGNKVISPSEDRKQPSNNLDRVKLTDFNFLMVLGKGSFGKVMLADRKGTEELYAIKILKKDVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKCA(5578)

Looking for similar products? Visit Product Comparison Guide

General description

PRKCA (protein kinase C, α) is a multi-domain signaling protein. It is predominantly expressed in the cytosol and nuclei. It is a Ca2+-dependent and phospholipid-dependent protein serine/threonine kinase. It is activated by diacylglycerol.

Immunogen

Protein kinase Cα-type recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRKCA (protein kinase C, α) is involved in the N-terminal phosphorylation of β-catenin. PKCα phosphorylates β-catenin by interacting directly at the Ser-Thr residue positions. It essentially performs in the Bcl2 (B-cell lymphoma 2) phosphorylation as well as suppression of apoptosis. During host defense and inflammation, it acts in LPS (lipopolysaccharide)-induced macrophage functions, controlling expression of tumor necrosis factor TNF-α , interleukin IL-1 secretion, and tumoricidal activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70021

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carter J Swanson et al.
The Journal of biological chemistry, 289(25), 17812-17829 (2014-05-03)
Signaling proteins comprised of modular domains have evolved along with multicellularity as a method to facilitate increasing intracellular bandwidth. The effects of intramolecular interactions between modular domains within the context of native proteins have been largely unexplored. Here we examine
Jungsug Gwak et al.
Biochemical and biophysical research communications, 450(4), 1673-1678 (2014-07-25)
Protein kinase Cα (PKCα) phosphorylates the Ser33/37/Thr41 residues of β-catenin, which lacks a typical PKCα canonical sequence, but little is known about its underlying mechanism. Here we showed that Ser33/Ser37/Thr41 of β-catenin fragments encompassing the armadillo repeats 1-5 (β-catenin1-781, β-catenin1-682
Lenka Dzurová et al.
Photodiagnosis and photodynamic therapy, 11(2), 213-226 (2014-03-04)
Hypericin photodynamic therapy (HypPDT) has been found to be an efficient inducer of cell death. However, there are indications that HypPDT also activates rescuing pathways. Cell responses to HypPDT are highly dependent on the Hyp intracellular localization and accumulation. We
A St-Denis et al.
The Journal of biological chemistry, 273(49), 32787-32792 (1998-11-26)
Lipopolysaccharide (LPS), a potent modulator of macrophage functional activity, binds to CD14 and triggers the activation of several protein kinases, leading to the secretion of variety of immunomodulatory molecules such as nitric oxide and proinflammatory cytokines. In this study, we
P P Ruvolo et al.
The Journal of biological chemistry, 273(39), 25436-25442 (1998-09-17)
Phosphorylation of Bcl2 at serine 70 may result from activation of a classic protein kinase C (PKC) isoform and is required for functional suppression of apoptosis by Bcl2 in murine growth factor-dependent cell lines (Ito, T., Deng, X., Carr, B.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service